Skip to Content
Merck
All Photos(1)

Key Documents

1.05707

Millipore

L-Methionine

for biochemistry

Synonym(s):

L-Methionine, Met

Sign Into View Organizational & Contract Pricing

Select a Size

1 KG
€501.00
10 KG
€2,840.00

€501.00


Estimated to ship on03 April 2025Details


Request a Bulk Order

Select a Size

Change View
1 KG
€501.00
10 KG
€2,840.00

About This Item

CAS Number:
Molecular Weight:
149.2
MDL number:
UNSPSC Code:
12352209
EC Index Number:
200-562-9
NACRES:
NA.21

€501.00


Estimated to ship on03 April 2025Details


Request a Bulk Order

Quality Level

form

solid

potency

36000 mg/kg LD50, oral (Rat)

pH

5-7 (20 °C, 10 g/L in H2O)

mp

280-282 °C

solubility

48 g/L

density

1.34 g/cm3

bulk density

550 kg/m3

storage temp.

2-30°C

SMILES string

S(CCC(N)C(=O)[O-])C.[H+]

InChI

1S/C5H11NO2S/c1-9-3-2-4(6)5(7)8/h4H,2-3,6H2,1H3,(H,7,8)

InChI key

FFEARJCKVFRZRR-UHFFFAOYSA-N

General description

for biochemistry

Application

L-Methionine for biochemistry. CAS 63-68-3, pH 5 - 7 (10 g/l, H₂O, 20 °C).

Analysis Note

Assay (perchloric acid titration, calculated on dried substance)≥ 99.0 %Identity (IR-spectrum)passes testAppearancewhite or almost white, crystalline powder or colorless crystalsAppearance of solution (0.5 mol/l, hydrochloric acid 1 mol/l)clear and colorlessSpec. rotation (α 20/D, 50 g/l, hydrochloric acid 1 mol/l, calc. on dried substance)+22.5 to +23.5UV-Absorption (260 nm, 0.5 mol/l, 1 cm, hydrochloric acid 1 mol/l)≤ 0.50UV-Absorption (280 nm, 0.5 mol/l, 1 cm, hydrochloric acid 1 mol/l)≤ 0.20Chloride (Cl)≤ 0.02 %Sulfate (SO₄)≤ 0.01 %Heavy metals (as Pb)≤ 0.001 %As (Arsenic)≤ 0.0005 %Ca (Calcium)≤ 0.001 %Co (Cobalt)≤ 0.0005 %Fe (Iron)≤ 0.0005 %K (Potassium)≤ 0.0005 %Mg (Magnesium)≤ 0.0005 %Na (Sodium)≤ 0.005 %NH₄ (Ammonium)≤ 0.01 %Zn (Zinc)≤ 0.0005 %Foreign amino acids≤ 0.3 %Other ninhydrine positive substances≤ 0.1 %Loss on drying (105 °C; 3 h)≤ 0.3 %

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service