Skip to Content
Merck
All Photos(2)

Key Documents

1.05321

SAFC

D(-)-Fructose

EMPROVE® ESSENTIAL, Ph. Eur., BP, USP, FCC

Pharma Manufacturing

Synonym(s):

D(-)-Fructose, Laevulose, Levulose

Sign Into View Organizational & Contract Pricing

Select a Size

25 KG
€792.00
21 X 25 KG
€22,000.00

€792.00


Available to ship on31 March 2025Details


Request a Bulk Order

Select a Size

Change View
25 KG
€792.00
21 X 25 KG
€22,000.00

About This Item

Empirical Formula (Hill Notation):
C6H12O6
CAS Number:
Molecular Weight:
180.16
MDL number:
UNSPSC Code:
12352201
EC Index Number:
200-333-3

€792.00


Available to ship on31 March 2025Details


Request a Bulk Order

Agency

BP
Ph. Eur.
USP

Quality Level

reg. compliance

FCC

description

EMPROVE® ESSENTIAL Ph Eur,BP,USP,FCC

product line

EMPROVE® ESSENTIAL

form

solid

autoignition temp.

360 °C

pH

5-6 (20 °C, 100 g/L in H2O)

mp

100-104 °C (decomposition)

solubility

790 g/L (OECD Test Guideline 105)

density

1.59 g/cm3 at 20 °C (OECD Test Guideline 109)

application(s)

liquid formulation
pharmaceutical
solid formulation

storage temp.

15-25°C

SMILES string

O[C@H]([C@H](O)CO)[C@H](O)C(=O)CO

InChI

1S/C6H12O6/c7-1-3(9)5(11)6(12)4(10)2-8/h3,5-9,11-12H,1-2H2/t3-,5-,6-/m1/s1

InChI key

BJHIKXHVCXFQLS-UYFOZJQFSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Finding the right excipient that matches your needs as well as regulatory demands can be a complicated challenge in formulation. With our application know-how and regulatory expertise, we support you in every step of development, scale-up, and production.
As part of our EMPROVE® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

Fructose is a commonly used ingredient in both food and pharmaceutics. In pharmaceutical application, fructose is used primarily for oral dosage forms.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service