Skip to Content
Merck
All Photos(1)

Key Documents

1.05272

Millipore

Antibiotic agar no. 1

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
€282.00

€282.00


Estimated to ship on07 April 2025Details


Request more information

Select a Size

Change View
500 G
€282.00

About This Item

UNSPSC Code:
41106213
NACRES:
NA.74

€282.00


Estimated to ship on07 April 2025Details


Request more information

Agency

AOAC®
EP
USP

Quality Level

reg. compliance

FDA

sterility

non-sterile

form

powder (dehydrated (DCM))

manufacturer/tradename

NutriSelect® Plus

technique(s)

microbe id | susceptibility testing: suitable
microbiological culture: suitable

pH

8.2-8.4 (25 °C in H2O)

application(s)

pharmaceutical

storage temp.

15-25°C

suitability

Micrococcus spp.
bacteria

Analysis Note

Appearance (clearness): clear
Appearance (color): yellowish-brown
pH-value (25 °C): 6.3 - 6.7
Solidification behaviour (2 hrs., 40 °C): liquid
Growth (Kocuria rhizophila ATCC 9341): good to very good
Inhibition zone diameter/Cephalotin 30 µg (Kocuria rhizophila ATCC 9341):
Inhibition zone diameter/Chloramphenicol 30 µg (Kocuria rhizophila ATCC 9341):
Inhibition zone diameter/Penicillin G 0.01 (Kocuria rhizophila ATCC 9341):
Inhibition zone diameter/Penicillin G 10 U/IE (Kocuria rhizophila ATCC 9341):
Growth (Staphylococcus aureus ATCC 6538-P (WDCM 00033)): good to very good
Inhibition zone diameter/Cephalotin 30 µg (Staphylococcus aureus ATCC 6538-P (WDCM 00033)):
Inhibition zone diameter/Chloramphenicol 30 µg (Staphylococcus aureus ATCC 6538-P (WDCM 00033)):
Inhibition zone diameter/Penicillin G 0.01 (Staphylococcus aureus ATCC 6538-P (WDCM 00033)):
Inhibition zone diameter/Penicillin G 10 U/IE (Staphylococcus aureus ATCC 6538-P (WDCM 00033)):
Growth (Bacillus subtilis ATCC 6633 (WDCM 00003)): good to very good
Growth (Staphylococcus epidermidis ATCC 12228 (WDCM 00036)): good to very good
Growth (Bacillus cereus ATCC 11778 (WDCM 00001)): good to very good
Incubation: 24 hrs.; 35°C; aerobic

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

AOAC is a registered trademark of AOAC International
GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service