Skip to Content
Merck
All Photos(1)

Key Documents

1.05194

Millipore

2-Oxoglutaric acid

for biochemistry

Synonym(s):

2-Oxoglutaric acid, 2-Ketoglutaric acid

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€53.70
100 G
€109.00
1 KG
€532.00
10 KG
€3,780.00

€53.70


Estimated to ship on30 April 2025Details


Request a Bulk Order

Select a Size

Change View
25 G
€53.70
100 G
€109.00
1 KG
€532.00
10 KG
€3,780.00

About This Item

CAS Number:
Molecular Weight:
146.09
MDL number:
UNSPSC Code:
12352106
EC Index Number:
206-330-3
NACRES:
NA.21

€53.70


Estimated to ship on30 April 2025Details


Request a Bulk Order

Quality Level

form

solid

pH

1.5 (20 °C, 50 g/L in H2O)

mp

112-116 °C

bulk density

800 kg/m3

storage temp.

2-8°C

SMILES string

[O-]C(=O)CCC(=O)C(=O)[O-].[H+].[H+]

InChI

1S/C5H6O5/c6-3(5(9)10)1-2-4(7)8/h1-2H2,(H,7,8)(H,9,10)

InChI key

KPGXRSRHYNQIFN-UHFFFAOYSA-N

General description

for biochemistry

Application

2-Oxoglutaric acid for biochemistry. CAS 328-50-7, pH 1.5 (50 g/l, H₂O, 20 °C).

Analysis Note

Assay99.0 - 101.0 %Identitypasses testMelting range112 - 115 °CAs (Arsenic)≤ 1 ppmFe (Iron)≤ 30 ppm

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service