Skip to Content
Merck
All Photos(1)

Key Documents

1.05178

Millipore

TBG broth, modified

Tetrathionate brilliant-green bile enrichment broth for microbiology

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
€282.00

€282.00


Estimated to ship on31 March 2025Details


Request more information

Select a Size

Change View
500 G
€282.00

About This Item

UNSPSC Code:
41106213
NACRES:
NA.74

€282.00


Estimated to ship on31 March 2025Details


Request more information

Quality Level

form

medium granules (dehydrated (DCM))

manufacturer/tradename

GranuCult®

technique(s)

microbiological culture: suitable

pH

7 (63 g/L in H2O, suspension)

application(s)

food and beverages
microbiology

storage temp.

15-25°C

suitability

Salmonella spp.

Analysis Note

Appearance (clearness): turbid with sediment
Appearance (colour): green
Inoculum on reference medium (Salmonella typhimurium ATCC 14028 (WDCM 00031)): 10 - 100
Inoculum on reference medium (Escherichia coli ATCC 25922 (WDCM 00013)): ≥ 1E+04
Reference: Tryptic soy agar
Enrichment from a mixed inoculum after 24 hours (Salmonella typhimurium ATCC 14028 (WDCM 00031)): ≥ 95 %
Enrichment from a mixed inoculum after 24 hours (Escherichia coli ATCC 25922 (WDCM 00013)): ≤ 5 %
Growth (Salmonella typhimurium ATCC 14028 (WDCM 00031)): +
Growth (Escherichia coli ATCC 25922 (WDCM 00013)): inhibited
Incubation: 24 hrs.; 35 °C; aerobic.
Detection on Brolacin agar.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. Tetrathionate Brilliant-green Bile enrichment broth, catalog # 105178. Can you send me a components list of this product. I was looking specifically if it contains Ox Bile. Thanks for your help.

    1 answer
    1. When prepared according to instructions, the Ox Bile concentration will be 8.5 g/L.

      Please see the link below to review the product datasheet, which includes the complete formulation on page 1:
      https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/312/639/tn7413en-ms.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service