Skip to Content
Merck

1.05020

SAFC

L-Leucine

EMPROVE® EXPERT, Ph. Eur., ChP, JP, USP

Pharma Manufacturing

Synonym(s):

L-Leucine, Leu

Sign Into View Organizational & Contract Pricing

Select a Size

1 KG
€936.00
1050209010
€2,955.00

€936.00


Estimated to ship on03 April 2025Details


Request a Bulk OrderRequest a Sample

Select a Size

Change View
1 KG
€936.00
1050209010
€2,955.00

About This Item

CAS Number:
Molecular Weight:
131.18
MDL number:
UNSPSC Code:
12352209
EC Index Number:
200-522-0

€936.00


Estimated to ship on03 April 2025Details


Request a Bulk OrderRequest a Sample

Agency

ChP
JP
Ph. Eur.
USP

Quality Level

vapor pressure

<1 hPa ( 20 °C)

description

EMPROVE® EXPERT Ph Eur,USP

product line

EMPROVE® EXPERT

pH

5.5-6.5 (20 °C, 20 g/L in H2O)

mp

270-340 °C (decomposition)

solubility

18.9 g/L

density

1.17 g/cm3 at 20 °C

bulk density

300‑400 kg/m3

application(s)

parenterals

storage temp.

2-30°C

SMILES string

NC(CC(C)C)C(=O)O

InChI

1S/C6H13NO2/c1-4(2)3-5(7)6(8)9/h4-5H,3,7H2,1-2H3,(H,8,9)

InChI key

ROHFNLRQFUQHCH-UHFFFAOYSA-N

General description

Our SAFC® portfolio of high-quality products for biopharmaceutical processing withstands strict quality control procedures and is produced according to MQ-500 requirements as defined by the M-Clarity program.

M-Clarity Program

As part of our EMPROVE® Program, our raw materials are offered with EMPROVE® Dossiers which provide comprehensive, up-to-date documentation to help you navigate regulatory challenges, manage risks, and improve your manufacturing processes.

Our comprehensive portfolio of upstream process chemicals not only provides biopharmaceutical manufacturers with high-quality raw materials for production of classical and novel therapies, but also helps them get to market faster and simplify regulatory challenges. Trust us to deliver supply chain transparency and reliable sourcing around the globe, streamlining your product qualification with best-in-class regulatory support and service.

Analysis Note

Assay (perchloric acid titration, calculated on dried substance)98.5 - 101.0 %Identity (IR-spectrum)passes testIdentity (specific rotation)passes testAppearancewhite to almost white, crystalline powder or shiny flakesAppearance of solution (50 g/l; hydrochloric acid 1 mol/l)clear and not more intense in color than reference solution BY₆Spec. rotation (α 20/D, 40 g/l, hydrochloric acid 250 g/l, calc. on dried substance)+14.5 to +16.5Spec. rotation (α 25/D, 40 g/l, hydrochloric acid 6 mol/l, calc. on dried substance)+14.9 to +17.3pH (10 g/l, CO₂-free water)5.5 - 7.0Chloride (Cl)≤ 200 ppmSulfate (SO₄)≤ 300 ppmFe (Iron)≤ 10 ppmFe (Iron) (ICP)≤ 10 ppmRelated Compounds, total (TLC)≤ 0.5 %Related compounds (HPLC) (Isoleucine)≤ 0.8 %Related compounds (HPLC) (any unspecified impurity)≤ 0.2 %Related compounds (HPLC) (total unspecified impurities)≤ 1.0 %Ninhydrin-positive substances (LC)(impurity A (570 nm))≤ 0.8 %Ninhydrin-positive substances (LC)(any ninhydrin-positive impurity)≤ 0.2 %Ninhydrin-positive substances (LC) (ammonium (570 nm))≤ 0.02 %Ninhydrin-positive substances (LC)(total impurities)≤ 1.0 %Residual solvents (ICH Q3C)excluded by manufacturing processSulfated ash (600 °C)≤ 0.1 %Loss on drying (105 °C; 3 h)≤ 0.2 %Bacterial endotoxins≤ 3.0 I.U./gTotal aerobic microbial count (TAMC)≤ 100 CFU/gTotal combined yeasts/moulds count (TYMC)≤ 100 CFU/gElemental impurity specifications have been set considering ICH Q3D (Guideline for Elemental Impurities). Class 1-3 elements are not likely to be present above the ICH Q3D option 1 limit, unless specified and indicated (*).Corresponds to Ph. Eur., USP

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


What is the Emprove® Program?

The Emprove® Program is a system providing comprehensive and thorough documentation of our filters and single-use components, pharma raw materials, and starting materials. Four document types are included with an Emprove® Program subscription:

Material Qualification Dossier

Free of charge - Sign in to download

Quality Management Dossier (Open Access)

Free of charge - Sign in to download

Operational Excellence Dossier

Available by subscription or for a fee

Learn more about the benefits an Emprove® Program subscription can provide
Why are these documents unavailable?
Emprove® Dossiers contain information intended to support the registration process and risk assessment activities of pharmaceutical producers and contain confidential information. Please contact us if you would like access.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service