Skip to Content
Merck
All Photos(1)

Key Documents

1.04885

Supelco

Potassium hydrogen sulfate

for analysis EMSURE® Reag. Ph Eur

Synonym(s):

Potassium hydrogen sulfate, Potassium bisulfate

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
€107.00
2.5 KG
€383.00
25 KG
€3,440.00

€107.00


Estimated to ship on03 March 2025Details


Request a Bulk Order

Select a Size

Change View
500 G
€107.00
2.5 KG
€383.00
25 KG
€3,440.00

About This Item

Empirical Formula (Hill Notation):
HKO4S
CAS Number:
Molecular Weight:
136.17
MDL number:
UNSPSC Code:
12352302
EC Index Number:
231-594-1
NACRES:
NA.21
Assay:
≥99.0% (alkalimetric)
form:
solid
solubility:
490 g/L (development of heat)

€107.00


Estimated to ship on03 March 2025Details


Request a Bulk Order

Agency

reag. Ph. Eur.

Quality Level

product line

EMSURE®

Assay

≥99.0% (alkalimetric)

form

solid

potency

2340 mg/kg LD50, oral (Rat)

pH

1 (20 °C, 50 g/L in H2O)

mp

195 °C (decomposition)

solubility

490 g/L (development of heat)

density

2.24 g/cm3 at 20 °C

bulk density

1140 kg/m3

anion traces

chloride (Cl-): ≤0.0005%
phosphate (PO43-): ≤0.001%

cation traces

Al: ≤0.002%
Ca: ≤0.005%
Fe: ≤0.0005%
N: ≤0.001%
Na: ≤0.005%
heavy metals (as Pb): ≤0.0005%

storage temp.

2-30°C

SMILES string

[K+].[K+].[S](=O)(=O)([O-])[O-]

InChI

1S/2K.H2O4S/c;;1-5(2,3)4/h;;(H2,1,2,3,4)/q2*+1;/p-2

InChI key

OTYBMLCTZGSZBG-UHFFFAOYSA-L

Related Categories

Analysis Note

Assay (alkalimetric): ≥ 99.0 %
Chloride (Cl): ≤ 0.0005 %
Phosphate (PO₄): ≤ 0.001 %
Total nitrogen (N): ≤ 0.001 %
Heavy metals (as Pb): ≤ 0.0005 %
Al (Aluminium): ≤ 0.002 %
Ca (Calcium): ≤ 0.005 %
Fe (Iron): ≤ 0.0005 %
Na (Sodium): ≤ 0.005 %
Corresponds to Reag. Ph Eur

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service