Skip to Content
Merck
All Photos(1)

Key Documents

1.04675

SAFC

Synthechol® Synthetic Cholesterol

non-GMP Sample

Synonym(s):

Cholesterol, 3β-Hydroxy-5-cholestene, 5-Cholesten-3β-ol

Sign Into View Organizational & Contract Pricing

Select a Size

1046750001
€405.00
1046750010
€2,025.00

€405.00


Estimated to ship on03 April 2025Details


Request a Bulk Order

Select a Size

Change View
1046750001
€405.00
1046750010
€2,025.00

About This Item

Empirical Formula (Hill Notation):
C27H46O
CAS Number:
Molecular Weight:
386.65
Beilstein:
1915888
MDL number:
UNSPSC Code:
12352212

€405.00


Estimated to ship on03 April 2025Details


Request a Bulk Order

biological source

synthetic

Quality Level

bp

360 °C (lit.)

mp

147-149 °C (lit.)

density

1.067 g/mL at 25 °C (lit.)

application(s)

advanced drug delivery
pharma/biopharma processes

storage temp.

no temp limit

SMILES string

CC(C)CCC[C@@H](C)[C@H]1CC[C@H]2[C@@H]3CC=C4C[C@@H](O)CC[C@]4(C)[C@H]3CC[C@]12C

InChI

1S/C27H46O/c1-18(2)7-6-8-19(3)23-11-12-24-22-10-9-20-17-21(28)13-15-26(20,4)25(22)14-16-27(23,24)5/h9,18-19,21-25,28H,6-8,10-17H2,1-5H3/t19-,21+,22+,23-,24+,25+,26+,27-/m1/s1

InChI key

HVYWMOMLDIMFJA-DPAQBDIFSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We offer a range of high quality critical raw materials plus the documentation and expertise to help our customers meet stringent regulatory requirements. Our SAFC® portfolio of high-quality products withstands strict quality control procedures.

Application

This product offers non-GMP samples representative of GMP material: Synthechol® Synthetic Cholesterol, EMPROVE® Expert; Ph. Eur., BP, JP, NF (104672).

Biochem/physiol Actions

Major component of all biological membranes; ~25% of total brain lipid is cholesterol.

Packaging

104675 0001 / 1G / PE Bag in Compound PE Inliner in PE Bottle
104675 0010 / 10G / PE Bag in Compound PE Inliner in PE Bottle

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany
SyntheChol is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service