1.04451
ICP Multi element standard solution XIII certified reference material
(15 elements in diluted nitric acid), Certipur®
Select a Size
€254.00
Estimated to ship on31 March 2025Details
Select a Size
About This Item
€254.00
Estimated to ship on31 March 2025Details
Recommended Products
Quality Level
matrix
(in diluted nitric acid)
pH
1 (20 °C in water)
density
1.032 g/cm3 at 20 °C
format
multi-component solution
storage temp.
15-25°C
General description
Please visit ISO certificates and Site Quality Self-Assessments to access the current certificates of accreditation.
Download your certificate at https://www.sigmaaldrich.com/US/en/documents-search?tab=coa to view certified values, including uncertainty and expiry date.
Application
Packaging
Analysis Note
As (arsenic): 100 mg/l
Be (beryllium): 100 mg/l
Cd (cadmium): 25 mg/l
Co (cobalt): 100 mg/l
Cr (chromium): 100 mg/l
Cu (copper): 100 mg/L
Fe (iron): 100 mg/l
Hg (mercury): 5 mg/l
Mn (managanese): 100 mg/l
Ni (nickel): 100 mg/l
Pb (lead): 100 mg/l
Se (selenium): 25 mg/l
V (vanadium): 250 mg/l
Zn (zinc): 100 mg/l
Other Notes
For detailed information for our newly launched ICP-Multi element Standards check the flyer: Commitment to Quality: Elevating Certipur® Standards to Certified Reference Material
In our effort to be more sustainable, we′re going digital with COAs-download them directly from our website.
Legal Information
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service