Skip to Content
Merck
All Photos(1)

Key Documents

1.04351

Millipore

L-Histidine

for biochemistry

Synonym(s):

His, (S)-2-Amino-3-(4-imidazolyl)propionic acid, NSC 137773

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€55.70
100 G
€118.00
1 KG
€835.00
10 KG
€4,220.00

€55.70


Estimated to ship on04 April 2025Details


Request a Bulk Order

Select a Size

Change View
25 G
€55.70
100 G
€118.00
1 KG
€835.00
10 KG
€4,220.00

About This Item

Empirical Formula (Hill Notation):
C6H9N3O2
CAS Number:
Molecular Weight:
155.15
Beilstein:
84088
MDL number:
UNSPSC Code:
12352209
EC Index Number:
200-745-3
NACRES:
NA.21

€55.70


Estimated to ship on04 April 2025Details


Request a Bulk Order

Quality Level

form

solid

potency

>5110 mg/kg LD50, oral (Rat)

pH

7.7 (20 °C, 10 g/L in H2O)

mp

282 °C (dec.) (lit.)

solubility

38.2 g/L

bulk density

430 kg/m3

storage temp.

2-30°C

SMILES string

N[C@@H](Cc1c[nH]cn1)C(O)=O

InChI

1S/C6H9N3O2/c7-5(6(10)11)1-4-2-8-3-9-4/h2-3,5H,1,7H2,(H,8,9)(H,10,11)/t5-/m0/s1

InChI key

HNDVDQJCIGZPNO-YFKPBYRVSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Precursor of histamine by action of histidine decarboxylase.

Analysis Note

Assay (acidimetric, calculated on dried substance): 99.0 - 101.0 %
Identity (IR-spectrum): passes test
Appearance: white to almost white, fine-crystalline powder or colorless crystals
Appearance of solution (50 g/l; water): clear and colorless
Spec. rotation (α 20/D, 110 g/l, hydrochloric acid 120 g/l, calc. on dried substance): +11.4 to +12.4
Chloride (Cl): ≤ 200 ppm
Sulfate (SO₄): ≤ 200 ppm
Heavy metals (as Pb): ≤ 10 ppm
As (Arsenic): ≤ 5 ppm
Ca (Calcium): ≤ 10 ppm
Co (Cobalt): ≤ 5 ppm
Fe (Iron): ≤ 5 ppm
K (Potassium): ≤ 5 ppm
Mg (Magnesium): ≤ 5 ppm
Na (Sodium): ≤ 100 ppm
Zn (Zinc): ≤ 5 ppm
Ninhydrin-positive substances (LC)(any ninhydrin-positive impurity): ≤ 0.2 %
Ninhydrin-positive substances (LC) (ammonium (570 nm)): ≤ 0.02 %
Ninhydrin-positive substances (LC)(total impurities): ≤ 0.5 %
Loss on drying (105 °C; 3 h): ≤ 0.2 %

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service