Skip to Content
Merck
All Photos(1)

Key Documents

1.03984

Millipore

Folic acid

for biochemistry

Synonym(s):

Folic acid, Pteroylglutamic acid

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€67.80
25 G
€139.00

€67.80


Estimated to ship on02 April 2025Details


Request a Bulk Order

Select a Size

Change View
5 G
€67.80
25 G
€139.00

About This Item

Empirical Formula (Hill Notation):
C19H19N7O6
CAS Number:
Molecular Weight:
441.40
MDL number:
UNSPSC Code:
12352106
EC Index Number:
200-419-0
NACRES:
NA.21

€67.80


Estimated to ship on02 April 2025Details


Request a Bulk Order

Quality Level

form

solid

pH

4.0-4.8 (25 °C, 100 g/L in H2O)

mp

250 °C

solubility

0.002 g/L

bulk density

110 kg/m3

storage temp.

15-25°C

SMILES string

[nH]1c2ncc(nc2[c](nc1N)=O)CNc3ccc(cc3)C(=O)N[C@@H](CCC(=O)O)C(=O)O

InChI

1S/C19H19N7O6/c20-19-25-15-14(17(30)26-19)23-11(8-22-15)7-21-10-3-1-9(2-4-10)16(29)24-12(18(31)32)5-6-13(27)28/h1-4,8,12,21H,5-7H2,(H,24,29)(H,27,28)(H,31,32)(H3,20,22,25,26,30)/t12-/m0/s1

InChI key

OVBPIULPVIDEAO-LBPRGKRZSA-N

General description

for biochemistry

Application

Folic acid for biochemistry. CAS 59-30-3, pH 4.0 - 4.8 (100 g/l, H₂O, 25 °C).

Analysis Note

Assay (HPLC, calc.on anhydrous substance)97.0 - 102.0 %Identity (HPLC)conformsIdentity (IR-spectrum)conformsAbsorption ratio (A₂₅₆/A₃₆₅)2.80 - 3.00Spec. rotation (α 20/D; 10 g/l; NaOH 0,1 mol/l; calc. on anhydrous subst.)+18 - +22 °Related substances (HPLC, Ph Eur) (impurity A)≤ 0.5 %Related substances (HPLC, Ph Eur) (impurity D)≤ 0.6 %Related compounds (HPLC) (total unspecified impurities)≤ 1.0 %Water (according to Karl Fischer)5.0 - 8.5 %

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service