Skip to Content
Merck
All Photos(1)

Key Documents

1.03604

Sigma-Aldrich

Diastase

(from fungi) for biochemistry

Synonym(s):

Diastase

Sign Into View Organizational & Contract Pricing

Select a Size

50 G
€104.00

€104.00


Estimated to ship on31 March 2025Details



Select a Size

Change View
50 G
€104.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352204
EC Index Number:
232-565-6
NACRES:
NA.21

€104.00


Estimated to ship on31 March 2025Details


Quality Level

form

solid

potency

>7500 mg/kg LD50, oral (Rat)

pH

6.5-7.5 (20 °C, 10 g/L in H2O)

solubility

10 g/L

bulk density

100‑200 kg/m3

storage temp.

2-8°C

Biochem/physiol Actions

Diastase is a mixture of α-and β-amylase and is found in honey. It mediates glycogen digestion and catalyzes the breakdown of starch to maltose. Diastase is used as a reducing and stabilizing agent in the aqueous synthesis of gold nanoparticles (AuNPs).

Analysis Note

Appearance: slightly brownish powder
α-Amylase (starch; pH 6.0; 25 °C): about 100
ß-Amylase(starch, pH 4.8; 25 °C): about 100
Note: Sulfatase and phosphatase detectable. Suitable for the determination of thiamine (Vit. B1) in food.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service