Skip to Content
Merck
All Photos(1)

Key Documents

1.02533

Supelco

Cobalt(II) chloride hexahydrate

extra pure

Synonym(s):

Cobalt dichloride, Cobaltous chloride hexahydrate

Sign Into View Organizational & Contract Pricing

Select a Size

1 KG
€530.00

€530.00


Available to ship on31 March 2025Details


Request a Bulk Order

Select a Size

Change View
1 KG
€530.00

About This Item

Linear Formula:
CoCl2 · 6H2O
CAS Number:
Molecular Weight:
237.93
MDL number:
UNSPSC Code:
12352302
NACRES:
NA.21
grade:
for analytical purposes

€530.00


Available to ship on31 March 2025Details


Request a Bulk Order

grade

for analytical purposes

Quality Level

vapor pressure

40 mmHg ( 0 °C)

pH

4.9 (25 °C, 50 g/L in H2O)

mp

56 °C

SMILES string

O.O.O.O.O.O.Cl[Co]Cl

InChI

1S/2ClH.Co.6H2O/h2*1H;;6*1H2/q;;+2;;;;;;/p-2

InChI key

GFHNAMRJFCEERV-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B Inhalation - Eye Dam. 1 - Muta. 2 - Repr. 1B - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

6.1D - Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)

CAS No.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service