Skip to Content
Merck
All Photos(1)

Key Documents

1.02487

Sigma-Aldrich

Chromium(III) chloride hexahydrate

Technipur®

Synonym(s):

Chromic chloride hexahydrate, Chromium trichloride hexahydrate, Hexaaquachromium(III) chloride, Trichlorochromium hexahydrate

Sign Into View Organizational & Contract Pricing

Select a Size

1 KG
€120.00
50 KG
€3,450.00

€120.00


Estimated to ship on31 March 2025Details


Request a Bulk Order

Select a Size

Change View
1 KG
€120.00
50 KG
€3,450.00

About This Item

Linear Formula:
CrCl3 · 6H2O
CAS Number:
Molecular Weight:
266.45
MDL number:
UNSPSC Code:
12352302
NACRES:
NA.21

€120.00


Estimated to ship on31 March 2025Details


Request a Bulk Order

Quality Level

pH

2.4-2.6 (20 °C, 50-53 g/L in H2O)

mp

81 °C

density

1.76 g/mL at 25 °C (lit.)

SMILES string

[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].Cl[Cr](Cl)Cl

InChI

1S/3ClH.Cr.6H2O/h3*1H;;6*1H2/q;;;+3;;;;;;/p-3

InChI key

LJAOOBNHPFKCDR-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Application

Chromium(III) chloride hexahydrate can be used as a:
  • Source of Cr(II) in the Takai reaction.[1]
  • Component of the deep eutectic solvent applicable as a catalyst in the esterification of carboxylic acids with alcohols.[2]
  • Precursor to prepare metal-oxide-doped amorphous titanium oxides, which are used as catalysts in the oxidative dehydrogenation of ethylbenzene.[3]

Legal Information

Technipur is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Met. Corr. 1 - Skin Sens. 1B

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amorphous porous MxTi mixed oxides as catalysts for the oxidative dehydrogenation of ethylbenzene
Li WY, et al.
Catalysis Letters, 69, 181?188 -181?188 (2000)
Deep eutectic solvent choline chloride?2CrCl3?6H2O: an efficient catalyst for esterification of formic and acetic acid at room temperature
Cao J, et al.
Royal Society of Chemistry Advances, 6, 21612-21616 (2016)
An Alternative Procedure in the Takai Reaction Using Chromium(III) Chloride Hexahydrate as a Convenient Source of Chromium(II)
Auge J, et al.
Synthetic Communications, 33(21) (2003)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service