Skip to Content
Merck
All Photos(1)

Key Documents

1.02330

Supelco

Cellulose microcrystalline

for thin-layer chromatography

Synonym(s):

Cellulose microcrystalline

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
€180.00

€180.00


Estimated to ship on09 April 2025Details


Request a Bulk Order

Select a Size

Change View
500 G
€180.00

About This Item

Linear Formula:
(C6H10O5)n
CAS Number:
MDL number:
UNSPSC Code:
41115711
EC Index Number:
232-674-9
NACRES:
NB.21

€180.00


Estimated to ship on09 April 2025Details


Request a Bulk Order

material

cellulose matrix
plastic support

Quality Level

form

microcrystalline

autoignition temp.

232 °C

potency

>5000 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rabbit)

feature

binder Organic Polymer
fluorescent indicator: no

packaging

pkg of 500 g

technique(s)

thin layer chromatography (TLC): suitable

particle size

5-40 μm

pore size

60 Å medium pore diameter

pH

5-7.5 (20 °C, 100 g/L in H2O, slurry)

transition temp

flash point 260 °C

density

1.5 g/cm3 at 20 °C

bulk density

70‑400 kg/m3

storage temp.

2-30°C

SMILES string

O1[C@H](C(C(C(C1CO)O)O)O)O[C@@H]2C(OC(C(C2O)O)O)CO

InChI

1S/C12H22O11/c13-1-3-5(15)6(16)9(19)12(22-3)23-10-4(2-14)21-11(20)8(18)7(10)17/h3-20H,1-2H2/t3?,4?,5?,6?,7?,8?,9?,10-,11?,12+/m1/s1

InChI key

GUBGYTABKSRVRQ-WFVLMXAXSA-N

General description

Standardized sorbents ensure reliable results every time. Our broad spectrum meets all kinds of needs. In TLC, silica gel 60 sorbent is considered the most versatile. Our assortment of silica gel 60 features different grades with particle sizes ranging from 5-40 µm: silica with a gypsum binder, silica with no foreign binder, and silica gel with a fluorescence indicator. We also offer high-quality aluminum oxide, cellulose microcrystalline, and kieselgur (also known as diatomaceous earth).

Application

  • Study of the influence of cellulose derivatives on physical and analytical attributes of a drug product belonging to BCS class II.: This research explored the impact of cellulose microcrystalline on the physical properties and analytical performance of a class II drug product. The findings demonstrated the role of cellulose microcrystalline in enhancing the stability and dissolution rate of the drug, contributing to improved bioavailability (Domoslawska et al., 2018).
  • Application of ethyl cellulose, microcrystalline cellulose and octadecanol for wax based floating solid dispersion pellets.: The study investigated the use of microcrystalline cellulose in the formulation of wax-based floating pellets. The results highlighted its effectiveness in controlling the release profile and enhancing the floating properties of the drug delivery system (Yan et al., 2016).
  • Modelling of porosity and waterfronts in cellulosic pellets for understanding drug release behavior.: This paper presented a model for analyzing the porosity and water penetration in cellulosic pellets, providing insights into the drug release mechanisms. The study emphasized the crucial role of microcrystalline cellulose in modulating drug release profiles (Gomez-Carracedo et al., 2010).
  • Fast dissolving films made of maltodextrins.: The research explored the incorporation of microcrystalline cellulose in fast-dissolving films, demonstrating its impact on film integrity and dissolution rates. The findings support the use of microcrystalline cellulose in enhancing the performance of oral thin films (Cilurzo et al., 2008).
  • Compressibility of floating pellets with verapamil hydrochloride coated with dispersion Kollicoat SR 30 D.: This study evaluated the compressibility and mechanical properties of floating pellets containing microcrystalline cellulose, showing its role in maintaining pellet integrity and controlled drug release (Sawicki & Lunio, 2005).

Analysis Note

d 10 (Laser diffraction, size distribution): 3.0 - 10.0 µm
d 50 (laser diffraction, size distribution): 10 - 30 µm
d 90 (Laser diffraction, size distribution): 30 - 60 µm

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

1–3 of 3 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  3. what is the molecular weight of microcrystalline cellulose for tlc

    1 answer
    1. The molecular weight for this product is not determined.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service