Skip to Content
Merck
All Photos(1)

Key Documents

1.02276

Supelco

Ammonium cerium(IV) nitrate

for analysis EMSURE® ACS,Reag. Ph Eur

Synonym(s):

Ammonium cerium(IV) nitrate, di-Ammonium-hexanitratocerate(IV), Cerium(IV) ammonium nitrate

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
€138.00
1 KG
€986.00

€138.00


Estimated to ship on03 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 G
€138.00
1 KG
€986.00

About This Item

Linear Formula:
(NH4)2[Ce(NO3)6]
CAS Number:
Molecular Weight:
548.22
MDL number:
UNSPSC Code:
12352302
EC Index Number:
240-827-6
NACRES:
NA.21
grade:
ACS reagent
form:
crystalline
solubility:
919-1020 g/L

€138.00


Estimated to ship on03 April 2025Details


Request a Bulk Order

grade

ACS reagent

Quality Level

Agency

reag. Ph. Eur.

product line

EMSURE®

Assay

≥99.0% ((NH4)2[Ce(NO3)6] basis, potentiometry)

form

crystalline

potency

300-2000 mg/kg LD50, oral (Rat)

impurities

≤0.005% Substances insoluble in diluted sulfuric acid

pH

0.61 (20 °C, 111.11 g/L in H2O)

mp

>400 °C

solubility

919-1020 g/L

density

2.49 g/cm3 at 24 °C

bulk density

1200 kg/m3

anion traces

chloride (Cl-): ≤0.001%
phosphate, silicate, arsenate (as PO43-): ≤0.005%

cation traces

Fe: ≤0.003%
Na: ≤0.005%
heavy metals (as Pb): ≤0.002%

storage temp.

2-30°C

SMILES string

[Ce+4].[N+](=O)([O-])O.[N+](=O)([O-])O.[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].N.N

InChI

1S/Ce.2HNO3.4NO3.2H3N/c;6*2-1(3)4;;/h;2*(H,2,3,4);;;;;2*1H3/q+4;;;4*-1;;

InChI key

WIBGOERAEYJBOT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application


  • A chemiluminescence method for screening of fluoroquinolones in milk samples based on a multi-pumping flow system.: This study presents a method for detecting fluoroquinolones in milk, using a multi-pumping flow system integrated with chemiluminescence detection facilitated by ammonium cerium(IV) nitrate. The research highlights the application of ammonium cerium(IV) nitrate in enhancing chemiluminescence and improving detection limits in food safety assays (Vakh et al., 2019).

Analysis Note

Assay (potentiometric, (NH₄)₂ [Ce(NO₃)₆!: ≥ 99.0 %
Substances insoluble in diluted sulfuric acid: ≤ 0.005 %
Chloride (Cl): ≤ 0.001 %
Phosphate (PO₄): ≤ 0.005 %
Heavy metals (as Pb): ≤ 0.002 %
Fe (Iron): ≤ 0.003 %
Na (Sodium): ≤ 0.005 %

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Met. Corr. 1 - Ox. Sol. 2 - Skin Corr. 1B - Skin Sens. 1A

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service