Skip to Content
Merck
All Photos(1)

Key Documents

1.02243

Sigma-Aldrich

Ethidium bromide adsorber

for decontamination of ethidium bromide from staining solutions

Sign Into View Organizational & Contract Pricing

Select a Size

1 UNIT
€92.20

€92.20


Available to ship on31 March 2025Details



Select a Size

Change View
1 UNIT
€92.20

About This Item

UNSPSC Code:
12352401
NACRES:
NA.51

€92.20


Available to ship on31 March 2025Details


pH

3.0 (25 °C in H2O, suspension)

storage temp.

15-25°C

Analysis Note

Performance test: passes test

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service