Skip to Content
Merck
All Photos(1)

Key Documents

1.02183

Supelco

Charcoal activated

pure

Synonym(s):

Charcoal activated

Sign Into View Organizational & Contract Pricing

Select a Size

1 KG
€112.00
1021839015
€545.00
17.5 KG
€782.00

€112.00


Estimated to ship on10 June 2025Details


Request a Bulk Order

Select a Size

Change View
1 KG
€112.00
1021839015
€545.00
17.5 KG
€782.00

About This Item

Empirical Formula (Hill Notation):
C
CAS Number:
Molecular Weight:
12.01
MDL number:
UNSPSC Code:
11101522
EC Index Number:
231-153-3
NACRES:
NB.21

€112.00


Estimated to ship on10 June 2025Details


Request a Bulk Order

Quality Level

form

solid

surface area

800-1500 m2/g

impurities

≤10% HNO3 solubles

ign. residue

≤10% (600 °C)

loss

≤10% loss on drying

mp

3550 °C

bulk density

150‑440 kg/m3

cation traces

Fe: ≤1000 ppm
Zn: ≤200 ppm

storage temp.

no temp limit

SMILES string

[C]

InChI

1S/C

InChI key

OKTJSMMVPCPJKN-UHFFFAOYSA-N

Analysis Note

Identity: passes test
Substances soluble in nitric acid: ≤ 10 %
Fe (Iron): ≤ 1000 ppm
Zn (Zinc): ≤ 200 ppm
n-Hexane adsorption: ≥ 30 %
Residue on ignition (600 °C): ≤ 10 %
Loss on drying: ≤ 10 %

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. What is the production technique for this material? Acid treatment with HNO3?

    1 answer
    1. This product is manufactured by the coking of natural beechwood. The activation process is considered proprietary.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service