Skip to Content
Merck
All Photos(1)

Key Documents

1.02111

Supelco

Calcium hydroxide

for the preparation of DIN 19266 pH standard buffer solutions

Synonym(s):

Calcium hydroxide

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€156.00

€156.00


Estimated to ship on31 March 2025Details


Request a Bulk Order

Select a Size

Change View
25 G
€156.00

About This Item

Linear Formula:
Ca(OH)2
CAS Number:
Molecular Weight:
74.09
MDL number:
UNSPSC Code:
12352305
EC Index Number:
215-137-3
NACRES:
NA.24

€156.00


Estimated to ship on31 March 2025Details


Request a Bulk Order

grade

analytical standard

Quality Level

Agency

complies with DIN 19266

potency

7340 mg/kg LD50, oral (Rat)

pH

12.6 (20 °C in H2O, saturated solution)

mp

550 °C (decomposition)

solubility

1.7 g/L

density

2.24 g/cm3 at 20 °C

bulk density

400 kg/m3

application(s)

environmental
food and beverages
general analytical
industrial qc
pharmaceutical

format

mixture

storage temp.

15-25°C

SMILES string

[Ca+2].[O-H].[O-H]

InChI

1S/Ca.2H2O/h;2*1H2/q+2;;/p-2

InChI key

AXCZMVOFGPJBDE-UHFFFAOYSA-L

General description

The pH buffer standards are measured and qualified using differential potentiometry with two platinum hydrogen electrodes. This method allows the comparison of the reference buffers to the corresponding primary reference materials by PTB (Physical Technical Institute, Germany) and NIST (National Institute of Standards and Technology, USA). Uncertainty data and traceability are documented in a comprehensive, batch-specific Certificate of Analysis.

Please visit ISO certificates and Site Quality Self-Assessments to access the current certificates of accreditation.

Download your certificate at http://www.sigma-aldrich.com to view certified values, including uncertainty and expiry date.

Application

For the preparation of DIN 19266 pH standard buffer solutions.

Calcium hydroxide (calcium dihydroxide or Ca(OH)2) is a reference buffer solution used to calibrate pH electrodes and pH instruments.

Analysis Note

pH-value (25°C): 12.43 - 12.47

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service