Skip to Content
Merck
All Photos(1)

Key Documents

1.02040

Sigma-Aldrich

Cesium carbonate

extra pure, Technipur®

Synonym(s):

Dicesium carbonate, Carbonic acid dicesium

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€46.50
1 KG
€1,190.00

€46.50


Estimated to ship on19 February 2025Details


Request a Bulk Order

Select a Size

Change View
25 G
€46.50
1 KG
€1,190.00

About This Item

Linear Formula:
Cs2CO3
CAS Number:
Molecular Weight:
325.82
Beilstein:
4546405
MDL number:
UNSPSC Code:
12352302
NACRES:
NA.21

€46.50


Estimated to ship on19 February 2025Details


Request a Bulk Order

Quality Level

pH

10-12 (20 °C, 50 g/L in H2O)

mp

610 °C (dec.) (lit.)

density

4.07 g/cm3 at 20 °C

SMILES string

[Cs+].[Cs+].[O-]C([O-])=O

InChI

1S/CH2O3.2Cs/c2-1(3)4;;/h(H2,2,3,4);;/q;2*+1/p-2

InChI key

FJDQFPXHSGXQBY-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Cesium carbonate (Cs2CO3) is widely used in electronic and optoelectronic industries because of its excellent electron injection capability and outstanding optical properties. It also possesses high solubility in polar solvents, therefore it can be used as a surface modification material in organic light-emitting diodes (OLEDs) and solar cells.[1][2]

Application

Cesium carbonate can be used as an additive in 4,7-diphenyl-1,10-phenanthroline (BPhen) based organic light-emitting diodes (OLEDs) to enhance the electrical characteristics and luminance efficiency of OLEDs by decreasing the activation energy of the organic layer as a result of increased electron injection and transport.[3][4] In perovskite solar cells (PCSs) and other optoelectronic devices, Cs2CO3 is used as an effective modifier to improve their photoelectric conversion efficiency by reducing the barrier between interfaces and defects in electron transport layers which makes electron transfer easier.[1][5]
Promotes the efficient O-alkylation of alcohols to form mixed alkyl carbonates.

Legal Information

Technipur is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Repr. 2 - STOT RE 2 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cesium carbonate modified electron transport layer for improving the photoelectric conversion efficiency of planar perovskite solar cells
Lv C, et al.
Materials Science in Semiconductor Processing, 112, 105014-105014 (2020)
The role of cesium carbonate on the electron injection and transport enhancement in organic layer by admittance spectroscopy
Zhao J, et al.
Applied Physics Letters, 101, 193303-193303 (2012)
Purification and preparation of some caesium compounds
Simons EL
Talanta, 13(2) (1966)
Development of an inorganic cesium carbonate-based electron transport material for a 17% power conversion efficiency perovskite solar cell device
Hossain MI, et al.
journal of photonics for energy, 10(1) (2020)
Improved open-circuit voltage via Cs2CO3-Doped TiO2 for high-performance and stable perovskite solar cells
Zong H, et al.
Organic Electronics, 77, 105495-105495 (2020)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service