Skip to Content
Merck
All Photos(1)

Key Documents

1.01963

Supelco

Potassium hydrogen tartrate

certified secondary standard reference material for pH measurement; directly traceable to primary SRM from NIST/PTB pH(S) = 3.63₉ / pH(S) = 3.55₇ (25°C) Certipur®

Synonym(s):

Potassium hydrogen tartrate, KHT, Monopotassium tartrate, Potassium Bitartrate, Potassium acid tartrate

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€193.00

€193.00


Estimated to ship on11 April 2025Details


Request a Bulk Order

Select a Size

Change View
25 G
€193.00

About This Item

Empirical Formula (Hill Notation):
C4H5KO6
CAS Number:
Molecular Weight:
188.18
MDL number:
UNSPSC Code:
41116107
EC Index Number:
212-769-1
NACRES:
NA.24

€193.00


Estimated to ship on11 April 2025Details


Request a Bulk Order

grade

certified reference material

Quality Level

Agency

complies with DIN 19266

product line

Certipur®

pH

3.4-3.7 (20 °C in H2O, saturated solution)

mp

250 °C (decomposition)

solubility

5.7 g/L

bulk density

720 kg/m3

application(s)

environmental
food and beverages
general analytical
industrial qc
pharmaceutical

format

neat

storage temp.

15-25°C

SMILES string

[K+].[O-]C(=O)[C@H](O)[C@@H](O)C(=O)O

InChI

1S/C4H6O6.K/c5-1(3(7)8)2(6)4(9)10;/h1-2,5-6H,(H,7,8)(H,9,10);/q;+1/p-1/t1-,2-;/m1./s1

InChI key

KYKNRZGSIGMXFH-ZVGUSBNCSA-M

General description

Certipur® certified secondary standard reference buffers are used to precisely calibrate and monitor pH measurement instruments and buffer solutions. They are quality controlled and qualified in our ISO/IEC 17025 and 17034 accredited calibration laboratories against certified primary standard reference materials (SRM) in compliance with DIN 19266, IUPAC, NIST and Ph. Eur. Certipur® reference materials offer maximum accuracy, reliability and convenience and help avoid costly repeat analyses.
Please visit ISO certificates and Site Quality Self-Assessments to access the current certificates of accreditation.
Download your certificate at http://www.sigma-aldrich.com to view certified values, including uncertainty and expiry date.

Application

Potassium hydrogen tartrate [pH(S) = 3.639 / pH(S) = 3.557 (25 °C)] is a certified secondary standard reference buffer used as a calibration buffer standard for pH instruments or pH electrodes.

Features and Benefits

  • The product is delivered with a comprehensive, batch-specific Certificate of Analysis showing relevant data for the quality management system.
  • The pH value of this Certipur® buffer solution is measured using differential potentiometry with two platinum hydrogen electrodes against NIST & PTB primary SRMs [pH(S) = 3.639 / pH(S) = 3.557 (25 °C)].
  • It is a Certified Reference Material (CRM) offered in neat format.
  • Intended for extremely precise calibration, qualification and monitoring of pH instruments and for testing other buffer solutions.

Analysis Note

pH-value (25 °C): 3.634 - 3.640
Identity: passes test

Legal Information

CERTIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

410.0 °F

Flash Point(C)

210 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service