Skip to Content
Merck
All Photos(1)

Documents

1.01546

Sigma-Aldrich

N,N′-Methylenebisacrylamide

for electrophoresis, special grade for molecular biology

Synonym(s):

N,N′-Methylenebisacrylamide, BIS, Bisacrylamide solution

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH2CHCONH)2CH2
CAS Number:
Molecular Weight:
154.17
MDL number:
UNSPSC Code:
41105319
EC Index Number:
203-750-9
NACRES:
NC.01

Assay

≥99.0% (saponification, acidimetric)

form

crystalline powder

potency

390 mg/kg LD50, oral (Rat)

technique(s)

electrophoresis: suitable

pH

≥5.0 (20 °C, 25 g/L in H2O)

mp

185 °C (decomposition)

density

1.235 g/cm3 at 30 °C

bulk density

200 kg/m3

storage temp.

2-8°C

InChI

1S/C7H10N2O2/c1-3-6(10)8-5-9-7(11)4-2/h3-4H,1-2,5H2,(H,8,10)(H,9,11)

InChI key

ZIUHHBKFKCYYJD-UHFFFAOYSA-N

Analysis Note

Appearance (colour): almost white
Appearance (description): crystalline powder
Free acid (as acrylic acid): ≤ 0.02 %
Water: ≤ 0.5 %
Assay (by acidimetry following saponification): ≥ 99.0 %
Identity (IR-spectrum): passes test
DNases (Exo- and endonucleases): not detectable
RNases: not detectable
Proteases: not detectable

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Carc. 1B - Muta. 1B - Repr. 2 - STOT RE 1 Oral

Target Organs

Peripheral nervous system

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service