Skip to Content
Merck
All Photos(1)

Key Documents

1.01307

Supelco

Titan yellow (C.I. 19540)

metal indicator, Reag. Ph Eur

Synonym(s):

Titan yellow (C.I. 19540), Clayton yellow, Naphthamine G, Thiazole yellow

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€73.20

€73.20


Estimated to ship on24 March 2025Details


Request a Bulk Order

Select a Size

Change View
25 G
€73.20

About This Item

Empirical Formula (Hill Notation):
C28H19N5Na2O6S4
CAS Number:
Molecular Weight:
695.72
MDL number:
UNSPSC Code:
12352107
EC Index Number:
217-377-4
NACRES:
NB.21

€73.20


Estimated to ship on24 March 2025Details


Request a Bulk Order

Product Name

Titan yellow (C.I. 19540), indicator Reag. Ph Eur

Agency

reag. Ph. Eur.

Quality Level

form

solid

loss

≤8% loss on drying, 110°C

pH

5.1 (20 °C, 10 g/L in H2O)

mp

>300 °C (decomposition)

solubility

29 g/L

bulk density

310 kg/m3

λmax

398-402 nm (buffer pH 7.0)

storage temp.

2-30°C

SMILES string

[Na+].[Na+].[S](=O)(=O)([O-])c1c2[s]c(nc2ccc1C)c3ccc(cc3)N\N=N\c4ccc(cc4)c5[s]c6c(n5)ccc(c6[S](=O)(=O)[O-])C

InChI

1S/C28H21N5O6S4.2Na/c1-15-3-13-21-23(25(15)42(34,35)36)40-27(29-21)17-5-9-19(10-6-17)31-33-32-20-11-7-18(8-12-20)28-30-22-14-4-16(2)26(24(22)41-28)43(37,38)39;;/h3-14H,1-2H3,(H,31,32)(H,34,35,36)(H,37,38,39);;/q;2*+1/p-2

InChI key

CZIRZNRQHFVCDZ-UHFFFAOYSA-L

Analysis Note

Identity (UV/VIS-Spectrum): passes test
Absorption maximum λmax. (buffer pH 7.0): 398 - 402 nm
Spec. Absorptivity A 1%/1cm (λmax; 0.01 g/l; buffer pH 7.0; calc. on dried substance): ≥ 350
Transition range: pH 12.0 - pH 13.0Yellow - orange
Loss on drying (110 °C): ≤ 8 %
Suitability as reagent (for magnesium): passes test

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service