Skip to Content
Merck
All Photos(3)

Documents

1.00996

Supelco

1-Propanol

EMPLURA®

Synonym(s):

1-Propanol, n-Propyl alcohol, Propan-1-ol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H8O
CAS Number:
Molecular Weight:
60.10
MDL number:
UNSPSC Code:
12191502
EC Index Number:
200-746-9
NACRES:
NA.21

vapor pressure

19 hPa ( 20 °C)

Quality Level

product line

EMPLURA®

Assay

≥99% (GC)

form

liquid

autoignition temp.

360 °C

expl. lim.

2.1-19.2 % (v/v)

impurities

≤0.002% Free acid (as propionic acid)
≤0.1% Methanol (GC)
≤0.2% Water

pH

7 (20 °C, 200 g/L in H2O)

bp

96.5-98 °C/1013 hPa

mp

-127 °C

transition temp

flash point 15 °C

density

0.80 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C3H8O/c1-2-3-4/h4H,2-3H2,1H3

InChI key

BDERNNFJNOPAEC-UHFFFAOYSA-N

General description

EMPLURA® is our low-cost alternative to high-purity qualities. With EMPLURA we offer a range of solvents for a plurality of basic applications in non-regulated industries and for less demanding applications, preparative laboratory work and cleaning purposes. EMPLURA solvents provide adequate specifications with the most important parameters.

Application


  • Noncontact In Situ Multidiagnostic NMR/Dielectric Spectroscopy: The study discusses the application of 1-Propanol in developing noncontact diagnostic techniques, beneficial for chemists and material scientists involved in analytical and diagnostic methodologies (Morais et al., 2024).

Analysis Note

Purity (GC): ≥ 99.0 %
Identity (IR): conforms
Free acid (as propionic acid): ≤ 0.002 %
Density (d 20 °C/ 4 °C): 0.803 - 0.804
Methanol (GC): ≤ 0.1 %
Readily carbonizable substances: conforms
Water: ≤ 0.20 %

Legal Information

EMPLURA is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

71.6 °F - closed cup

Flash Point(C)

22 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service