Skip to Content
Merck
All Photos(1)

Key Documents

1.00961

Supelco

Total Hardness Test, photometric

photometric, 5-215 mg/L (Ca), water hardness 0.7-30.1 °d, water hardness 0.9-37.6 °e, water hardness 1.2-53.7 °f, 7-301 mg/L (CaO), 12-537 mg/L (CaCO3), Spectroquant®

Sign Into View Organizational & Contract Pricing

Select a Size

25 TESTS
€163.00

€163.00


Estimated to ship on03 February 2025Details


Request a Bulk OrderRequest more information

Select a Size

Change View
25 TESTS
€163.00

About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

€163.00


Estimated to ship on03 February 2025Details


Request a Bulk OrderRequest more information

Product Name

Total Hardness Cell Test, photometric, 5-215 mg/L (Ca), water hardness 0.7-30.1 °d, water hardness 0.9-37.6 °e, water hardness 1.2-53.7 °f, 7-301 mg/L (CaO), 12-537 mg/L (CaCO3), Spectroquant®

product line

Spectroquant®

Quality Level

usage

sufficient for 25 tests

specific analyte(s)

calcium

measuring range

12-537 mg/L (CaCO3)
5-215 mg/L (Ca)
7-301 mg/L (CaO)

water hardness measuring range

0.7-30.1 °d
0.9-37.6 °e
1.2-53.7 °f

technique(s)

photometry: suitable

compatibility

for use with Spectroquant® Move 100
for use with Spectroquant® Nova 60 A
for use with Spectroquant® Prove 100
for use with Spectroquant® Prove 300
for use with Spectroquant® Prove 600

detection method

photometric (Phthalein complexone)

storage temp.

15-25°C

General description

Method: Photometric, 5 - 215 mg/L (Ca), 0.7 - 30.1 °d, 0.9 - 37.6 °e, 1.2 - 53.7 °f 7 - 301 mg/L (CaO), 12 - 537 mg/L (CaCO3), Spectroquant®

This Spectroquant® Total Hardness Cell Test allows the accurate quantification of the hardness of water samples. The total hardness of a given sample is due to its content of salts of the alkaline earth metals calcium, magnesium, strontium, and barium (“hardening constituents”).
Since strontium and barium are generally present in waters only in traces, the hardness is defined as the content in a water of calcium ions, Ca2+, and magnesium ions, Mg2+ (“hardness ions”).

Method applied: In neutral solution calcium and magnesium ions form with phthalein purple a violet dye that is determined photometrically.

The Spectroquant® Cell Tests come with prefilled 16 mm round cells and all the required reagents to perform the analysis according to the instruction leaflet provided.

All Spectroquant® Cell and Reagent Test Kits are equipped with the unique Live ID (2D barcode) which allows seamless method recognition and contains essential information such as lot number, expiry date, and automatic calibration updates.

Legal Information

SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

FlameHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 3 - Repr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service