Skip to Content
Merck
All Photos(2)

Key Documents

1.00918

Millipore

Sabouraud Dextrose Agar

GranuCult® plus, with Dextrose 4 %, Chloramphenicol, EP, USP, JP, granular, pkg of 500 g

Synonym(s):

SDA, Sabouraud Agar, Sabouraud Agar with Chloramphenicol

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
€243.00

€243.00


Estimated to ship on01 April 2025Details



Select a Size

Change View
500 G
€243.00

About This Item

UNSPSC Code:
41106231
NACRES:
NA.74

€243.00


Estimated to ship on01 April 2025Details


Agency

EP
JP
USP

Quality Level

form

granular

composition

Chloramphenicol
with Dextrose, 4%

packaging

pkg of 500 g

manufacturer/tradename

GranuCult® plus

technique(s)

microbiological culture: suitable

color

beige Dehydrated medium
clear yellowish to brownish Prepared medium

pH

5.6±0.2 (25 °C)

application(s)

cosmetics
food and beverages
pharmaceutical

storage temp.

15-25°C

suitability

nonselective for molds
nonselective for yeasts

Looking for similar products? Visit Product Comparison Guide

General description

Sabouraud Agar with Chloramphenicol is a nutrient rich complex media designed to promote the growth of yeast and molds. It is often used with antibiotics for the isolation of pathogenic fungi from samples containing large numbers of other fungi or bacteria. Sabouraud agar contains digests of animal tissues (peptones), which provide a nutritious source of amino acids and nitrogenous compounds for the growth of fungi and yeasts.
The high concentration of dextrose in addition with the low pH promotes the growth, the formation of spores (Conidia and Sporangia) as well as the formation of pigments of yeasts and molds. On the other side, growth of bacteria is inhibited.
The low pH favors fungal growth. The medium is supplemented with chloramphenicol to prevent the growth of associated bacteria present in the test sample.

Application

Sabouraud Agar with Chloramphenicol is used to cultivate yeast and molds from test samples. It also promotes the formation of spores (conidia and sporangia) and pigments of yeasts and molds. This media is suitable for microbiological examination of non-sterile products.

Storage and Stability

  • The product can be used for sampling until the expiry date if stored upright, protected from light, and properly sealed at +15 °C to +25 °C.
  • After the first opening of the bottle, the content can be used up to the expiry date when stored dry and tightly closed at +15 °C to +25 °C.
  • Prepared plates can be stored in sealed plastic pouches or bags for up to 2 weeks at 2 – 8 °C and protected from light.

Other Notes

There should be no visible moisture on the plates before use. When moisture is present, the plates should be dried for the minimum time required to remove visible moisture.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service