Skip to Content
Merck
All Photos(1)

Key Documents

1.00910

Supelco

Petroleum benzine boiling range 50-70°C

EMPLURA®

Synonym(s):

Petroleum ether

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12191502
NACRES:
NB.21
bp:
50-70 °C/1013 hPa
vapor pressure:
19-50 kPa ( 25 °C)
To inquire about this product 1.00910, please contact your local Merck office or dealer. Contact Technical Service

vapor pressure

19-50 kPa ( 25 °C)

Quality Level

product line

EMPLURA®

autoignition temp.

>200 °C

potency

16750 mg/kg LD50, oral (Rat)
3350 mg/kg LD50, skin (Rabbit)

expl. lim.

1.2-8.3 % (v/v)

dilution

(low-cost alternative, for basic applications)

impurities

≤0.0005% Free acid (as CH3COOH)
≤0.01% Water

evapn. residue

≤0.001%

kinematic viscosity

0.4-0.7 cSt(20 °C)

bp

50-70 °C/1013 hPa

mp

<-20 °C

transition temp

flash point -20 °C

density

0.65-0.7 g/cm3 at 15 °C

storage temp.

2-30°C

Looking for similar products? Visit Product Comparison Guide

General description

EMPLURA® is our low-cost alternative to high-purity qualities. With EMPLURA we offer a range of solvents for a plurality of basic applications in non-regulated industries and for less demanding applications, preparative laboratory work and cleaning purposes. EMPLURA solvents provide adequate specifications with the most important parameters.

Analysis Note

Free acid (as CH₃COOH): ≤ 0.0005 %
Boiling range (50-70°C): ≥ 90 % (v/v)
Density (d 20 °C/ 4 °C): 0.654 - 0.670
Readily carbonizable substances: conforms
Evaporation residue: ≤ 0.001 %
Water: ≤ 0.01 %

Legal Information

EMPLURA is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-4.0 °F

Flash Point(C)

-20 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service