Skip to Content
Merck
All Photos(1)

Key Documents

1.00678

Supelco

AOX Enrichment Set

Spectroquant®

Sign Into View Organizational & Contract Pricing

Select a Size

1 SET
€134.00

€134.00


Estimated to ship on24 March 2025Details


Request a Bulk OrderRequest more information

Select a Size

Change View
1 SET
€134.00

About This Item

UNSPSC Code:
41116116
NACRES:
SE.11

€134.00


Estimated to ship on24 March 2025Details


Request a Bulk OrderRequest more information

product line

Spectroquant®

Quality Level

analyte functional class(es)

adsorbable organic halides (AOX)

technique(s)

photometry: suitable

storage temp.

2-30°C

General description

This AOX Enrichment Set is a required accessory when performing the Spectroquant® AOX Cell Test Kit 1.00675. It contains 2 glass reservoirs for AOX enrichment.

Legal Information

SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service