Skip to Content
Merck
All Photos(1)

Key Documents

1.00354

Millipore

Indole-3-butyric acid

LAB

Synonym(s):

Indole-3-butyric acid, JBAA, IBA

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€57.20
25 G
€134.00
100 G
€380.00
500 G
€1,520.00

€57.20


Estimated to ship on02 April 2025Details


Request a Bulk Order

Select a Size

Change View
5 G
€57.20
25 G
€134.00
100 G
€380.00
500 G
€1,520.00

About This Item

Empirical Formula (Hill Notation):
C12H13NO2
CAS Number:
Molecular Weight:
203.24
MDL number:
UNSPSC Code:
12352106
EC Index Number:
205-101-5
NACRES:
NA.21

€57.20


Estimated to ship on02 April 2025Details


Request a Bulk Order

Quality Level

Assay

≥98.0% (acidimetric)

form

solid

mp

121-126 °C

solubility

0.25 g/L

bulk density

360 kg/m3

storage temp.

2-30°C

SMILES string

[nH]1c2c(c(c1)CCCC(=O)O)cccc2

InChI

1S/C12H13NO2/c14-12(15)7-3-4-9-8-13-11-6-2-1-5-10(9)11/h1-2,5-6,8,13H,3-4,7H2,(H,14,15)

InChI key

JTEDVYBZBROSJT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Octanal Enhances Disease Resistance in Postharvest Citrus Fruit by the Biosynthesis and Metabolism of Aromatic Amino Acids.: This paper discusses the indirect role of IBA in enhancing disease resistance in citrus fruits postharvest by facilitating the biosynthesis of aromatic amino acids. (Duan et al., 2024).

  • Callus Induction Followed by Regeneration and Hairy Root Induction in Common Buckwheat.: This research demonstrates the use of IBA for callus induction and subsequent regeneration and hairy root induction in common buckwheat, providing a valuable protocol for genetic studies and crop improvement. (Omelchenko et al., 2024).

Analysis Note

Assay (acidimetric): ≥ 98.0 %
Identity (IR-spectrum): passes test
Melting point: 122 - 125 °C

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service