Skip to Content
Merck
All Photos(1)

Key Documents

1.00345

Supelco

Hydroiodic acid

67%, for analysis EMSURE® ACS,ISO,Reag. Ph Eur

Synonym(s):

Hydrogen iodide solution

Sign Into View Organizational & Contract Pricing

Select a Size

250 ML
€365.00

€365.00


Estimated to ship on25 March 2025Details


Request a Bulk Order

Select a Size

Change View
250 ML
€365.00

About This Item

UNSPSC Code:
12352301
NACRES:
NA.21

€365.00


Estimated to ship on25 March 2025Details


Request a Bulk Order

Product Name

Hydroiodic acid 67%, for analysis EMSURE®

grade

reagent

Quality Level

product line

EMSURE®

Assay

≥67% (acidimetric)

form

liquid

impurities

≤0.1 ppm Ba (Barium)
≤0.1 ppm Be (Beryllium)
≤0.1 ppm Bi (Bismuth)
≤0.1 ppm Cd (Cadmium)
≤0.1 ppm Co (Cobalt)
≤0.1 ppm Cr (Chromium)
≤0.1 ppm Cu (Copper)
≤0.1 ppm Ge (Germanium)
≤0.1 ppm Li (Lithium)
≤0.1 ppm Mn (Manganese)
≤0.1 ppm Mo (Molybdenum)
≤0.1 ppm Ni (Nickel)
≤0.1 ppm Pb (Lead)
≤0.1 ppm Sr (Strontium)
≤0.1 ppm Ti (Titanium)
≤0.1 ppm Tl (Thallium)
≤0.1 ppm V (Vanadium)
≤0.1 ppm Zr (Zirconium)
≤0.5 ppm Zn (Zinc)
≤1 ppm Al (Aluminium)
≤1 ppm As (Arsenic)
≤1 ppm Ca (Calcium)
≤1 ppm K (Potassium)
≤1 ppm Na (Sodium)
≤10 ppm Phosphorous compounds (as PO4)
≤2 ppm Fe (Iron)
≤20 ppm Sulfate (SO4)
≤200 ppm Chloride and Bromide (as Cl)
≤5 ppm Mg (Magnesium)

ign. residue

≤50 ppm (as sulfate)

bp

127 °C/1013 hPa

density

1.97 g/cm3 at 20 °C

storage temp.

2-30°C

Related Categories

Analysis Note

Assay (acidimetric): ≥ 67 %
Chloride and Bromide (as Cl): ≤ 200 ppm
Phosphorous compounds (as PO₄): ≤ 10 ppm
Sulfate (SO₄): ≤ 20 ppm
Al (Aluminium): ≤ 1 ppm
As (Arsenic): ≤ 1 ppm
Ba (Barium): ≤ 0.1 ppm
Be (Beryllium): ≤ 0.1 ppm
Bi (Bismuth): ≤ 0.1 ppm
Ca (Calcium): ≤ 1 ppm
Cd (Cadmium): ≤ 0.1 ppm
Co (Cobalt): ≤ 0.1 ppm
Cr (Chromium): ≤ 0.1 ppm
Cu (Copper): ≤ 0.1 ppm
Fe (Iron): ≤ 2 ppm
Ge (Germanium): ≤ 0.1 ppm
K (Potassium): ≤ 1 ppm
Li (Lithium): ≤ 0.1 ppm
Mg (Magnesium): ≤ 5 ppm
Mn (Manganese): ≤ 0.1 ppm
Mo (Molybdenum): ≤ 0.1 ppm
Na (Sodium): ≤ 1 ppm
Ni (Nickel): ≤ 0.1 ppm
Pb (Lead): ≤ 0.1 ppm
Sr (Strontium): ≤ 0.1 ppm
Ti (Titanium): ≤ 0.1 ppm
Tl (Thallium): ≤ 0.1 ppm
V (Vanadium): ≤ 0.1 ppm
Zn (Zinc): ≤ 0.5 ppm
Zr (Zirconium): ≤ 0.1 ppm
Alkoxygroups: passes test
Residue on ignition (as sulfate): ≤ 50 ppm

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service