Skip to Content
Merck
All Photos(2)

Key Documents

07-748

Sigma-Aldrich

Anti-monomethyl-Histone H4 (Lys20) Antibody

Upstate®, from rabbit

Synonym(s):

H4K20me1, Histone H4 (mono methyl K20)

Sign Into View Organizational & Contract Pricing

Select a Size

200 μG
€495.00

€495.00


Estimated to ship on28 April 2025Details


Request a Bulk Order

Select a Size

Change View
200 μG
€495.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€495.00


Estimated to ship on28 April 2025Details


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

manufacturer/tradename

Upstate®

technique(s)

immunocytochemistry: suitable
multiplexing: suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

monomethylation (Lys20)

Gene Information

Specificity

Broad species cross-reactivity expected due to sequence conservation
monomethylated Histone H4 (Lys20)

Immunogen

KLH-conjugated, branched synthetic peptide corresponding to amino acids 15-25 (AKRHR[me1K]VLRDN) of human Histone H4

Application

Additional Referenced Research Applications:
Immunocytochemistry (cells)
Anti-monomethyl-Histone H4 (Lys20) Antibody is a rabbit polyclonal antibody for detection of monomethyl-Histone H4 (Lys20) also known as H4K20me1, Histone H4 (mono methyl K20) & has been validated in WB, Mplex, ICC.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Histones

Quality

Routinely evaluated by immunoblot

Target description

10kDa

Physical form

0.1M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide and 30% glycerol.
Format: Purified
Protein A purified

Storage and Stability

2 years at -20°C from date of shipment

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Li Liu et al.
PloS one, 8(11), e78253-e78253 (2013-11-14)
The upstream Gγ-globin cAMP-response element (G-CRE) plays an important role in regulating Gγ-globin expression through binding of ATF2 and its DNA-binding partners defined in this study. ATF2 knockdown resulted in a significant reduction of γ-globin expression accompanied by decreased ATF2

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service