Skip to Content
Merck
All Photos(1)

Key Documents

07-448-S

Sigma-Aldrich

Anti-monomethyl-Histone H3 (Lys27) Antibody, Trial Size

Upstate®, from rabbit

Synonym(s):

H3K27me1, Histone H3 (monomethyl K27), H3 histone family, member T, Histone 3, H3, Histone cluster 3, H3

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
€125.00

€125.00


Estimated to ship on11 March 2025Details


Request a Bulk Order

Select a Size

Change View
20 μG
€125.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€125.00


Estimated to ship on11 March 2025Details


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, vertebrates

manufacturer/tradename

Upstate®

technique(s)

ChIP: suitable (ChIP-seq)
dot blot: suitable
immunocytochemistry: suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

monomethylation (Lys27)

Gene Information

human ... HIST1H3F(8968)

General description

Histone H3 is one of the five main histone proteins involved in the structure of chromatin in eukaryotic cells. Featuring a main globular domain and a long N-terminal tail, H3 is involved with the structure of the nucleosomes of the ′beads on a string′ structure. The N-terminal tail of histone H3 protrudes from the globular nucleosome core and can undergo several different types of epigenetic modifications that influence cellular processes. These modifications include the covalent attachment of methyl or acetyl groups to lysine and arginine amino acids and the phosphorylation of serine or threonine.

Specificity

Broad species cross-reactivity expected, based on sequence homology.
Recognizes monomethylated histone H3 (Lys27), Mr 17 kDa.

Immunogen

Epitope: Lys27
KLH-conjugated, synthetic 2X-branched peptide containing the sequence …ARmeKSA… in which meK corresponds to monomethyl-lysine at residue 27 of human histone H3.

Application

Anti-monomethyl-Histone H3 (Lys27), Trial Size Antibody is a Rabbit Polyclonal Antibody for detection of Histone H3 monomethylated on lysine 27 (H3K27me1). This antibody has dot blot (DB) verified specificity and is validated in, ICC, WB, ChIP, ChIP-seq. Small pack size is ideal for screening.
Immunocytochemistry & Dot Blot:
Reported by an independent laboratory (Kohlmaier, A., 2004; Perez-Burgos, L., 2004; Peters, A. H., 2003).
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Histones

Quality

Routinely evaluated by western blot acid in extracted proteins from Hela cells.

Western Blot Analysis:
0.1-0.2 μg/mL had detected monomethylated histone H3 in acid extracted proteins from HeLa cells.

Target description

17 kDa

Physical form

Format: Purified
Protein A purified
Purified rabbit IgG in buffer containing 0.01 M phosphate buffer, pH 7.6, 0.125 M NaCl, 0.05% sodium azide and 30% glycerol.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
HeLa acid extract

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thea A Egelhofer et al.
Nature structural & molecular biology, 18(1), 91-93 (2010-12-07)
We have tested the specificity and utility of more than 200 antibodies raised against 57 different histone modifications in Drosophila melanogaster, Caenorhabditis elegans and human cells. Although most antibodies performed well, more than 25% failed specificity tests by dot blot
Ashley B Reers et al.
Epigenetics & chromatin, 16(1), 25-25 (2023-06-16)
Gene expression in malaria parasites is subject to various layers of regulation, including histone post-translational modifications (PTMs). Gene regulatory mechanisms have been extensively studied during the main developmental stages of Plasmodium parasites inside erythrocytes, from the ring stage following invasion

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service