Skip to Content
Merck
All Photos(1)

Key Documents

07-248

Sigma-Aldrich

Anti-BSA (Bovine Serum Albumin) Antibody

Upstate®, from rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
€153.00
200 μG
€622.00

€153.00


In StockDetails


Request a Bulk Order

Select a Size

Change View
25 μG
€153.00
200 μG
€622.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€153.00


In StockDetails


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse, bovine, human, horse, rat

packaging

antibody small pack of 25 μg

manufacturer/tradename

Upstate®

technique(s)

western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

bovine ... Alb(280717)
horse ... Alb(100034206)
human ... ALB(213)
mouse ... Alb(11657)
rat ... Alb(24186)

Specificity

BSA

Immunogen

Fraction V bovine serum albumin

Application

Research Category
Epitope Tags & General Use
Research Sub Category
Epitope Tags
This Anti-Albumin Antibody is an antibody against BSA (Bovine Serum Albumin) for use in WB.

Quality

routinely evaluated by immunoblot on BSA and albumin as a contaminant in lysates derived from various in vitro cultured cell lines

Target description

66kDa

Physical form

0.1M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide before the addition of glycerol to 30%
Format: Purified
Protein A chromatography

Storage and Stability

2 years at -20°C

Other Notes

Please refer to lot specific datasheet.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sugapriya Dhanasekaran et al.
Analytical cellular pathology (Amsterdam), 2015, 219206-219206 (2015-11-12)
The success of liver regeneration depends on the availability of suitable cell types and their potential to differentiate into functional hepatocytes. To identify the stem cells which have the ability to differentiate into hepatocytes, we used neonatal liver as source.
Thomas Simon et al.
International journal of molecular sciences, 21(9) (2020-05-01)
Extracellular vesicles (EVs) are nanosized structures able to carry proteins, lipids and genetic material from one cell to another with critical implications in intercellular communication mechanisms. Even though the rapidly growing EVs research field has sparked great interest in the
Kimberly J Reidy et al.
Development (Cambridge, England), 136(23), 3979-3989 (2009-11-13)
Semaphorin3a (Sema3a), a chemorepellant guidance protein, plays crucial roles in neural, cardiac and peripheral vascular patterning. Sema3a is expressed in the developing nephron, mature podocytes and collecting tubules. Sema3a acts as a negative regulator of ureteric bud branching, but its
Kimberly J Reidy et al.
The American journal of pathology, 183(4), 1156-1168 (2013-08-21)
Semaphorin-3A (Sema3a), a guidance protein secreted by podocytes, is essential for normal kidney patterning and glomerular filtration barrier development. Here, we report that podocyte-specific Sema3a gain-of-function in adult mice leads to proteinuric glomerular disease involving the three layers of the
Delma Veron et al.
PloS one, 7(7), e40589-e40589 (2012-07-19)
Podocyte or endothelial cell VEGF-A knockout causes thrombotic microangiopathy in adult mice. To study the mechanism involved in acute and local injury caused by low podocyte VEGF-A we developed an inducible, podocyte-specific VEGF-A knockdown mouse, and we generated an immortalized

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service