Skip to Content
Merck
All Photos(5)

Key Documents

06-913-I

Sigma-Aldrich

Anti-Sin3A Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Paired amphipathic helix protein Sin3a, Histone deacetylase complex subunit Sin3a, Transcriptional corepressor Sin3a, Sin3A

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€365.00

€365.00


Estimated to ship on23 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
€365.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€365.00


Estimated to ship on23 April 2025Details


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, mouse

species reactivity (predicted by homology)

rat (based on 100% sequence homology), rhesus macaque (based on 100% sequence homology)

technique(s)

immunocytochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... SAP30(8819)
mouse ... Sin3A(20466)

General description

Paired amphipathic helix protein Sin3a (UniProt Q60520; also known as Histone deacetylase complex subunit Sin3a, mSin3a, Transcriptional co-repressor Sin3a) is encoded by the Sin3A (also known as mSin3A, AW553200, Kiaa4126, mKIAA4126) gene (Gene ID 8819) in murine species. The transcription regulator Sin3a is a scaffold protein that interacts with various proteins to form specialized complexes that mediate different biological processes, including chromatin modification, embryonic development, DNA repair, mitochondrial metabolism, and gene transcription. SIN3a harbors four paired α-helices (PAH), a histone interaction domain, and a highly conserved region. The histone interaction domain mediates HDACs binding, whereas the highly conserved region interacts with proteins necessary for repression, such as the nuclear receptor hormone co-repressor Alien. The four PAHs represent the main interacting domains of SIN3a and mediate the interactions with a wide variety of DNA-binding transcription factors. Members of the Mad family of repressor proteins, for example, specifically interact with the PAH2 through their Sin3 interacting domain (SID). Although widely studied for its involvement in transcriptional repression, Sin3a is shown to be essential for aryl hydrocarbon receptor-mediated induction of gene transcription.

Immunogen

Epitope: N-terminus
KLH-conjugated peptide (KRRLDDQESPVYAAQQRR-C) corresponding to amino acids 2-19 of mouse Sin3A.

Application

Anti-Sin3A Antibody is an antibody against Sin3A Antibody for use in Western Blotting and Immunocytochemistry.
Immunocytochemistry Analysis: 4.0 µg/mL from a representative lot detected Sin3A in HeLa, A431, HUVEC, and NIH/3T3 cells.
Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional

Quality

Evaluated by Western Blotting in HeLa nuclear extract.

Western Blotting Analysis: 0.05 µg/mL of this antibody detected Sin3A in 10 µg of HeLa nuclear extract.

Target description

~160 kDa observed

Linkage

Replaces: 06-913

Physical form

Affinity purified
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service