Skip to Content
Merck
All Photos(1)

Key Documents

06-1334

Sigma-Aldrich

Anti-phospho-TAB1 (Thr431) Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

TGF-beta activated kinase 1/MAP3K7 binding protein 1, TAK1-binding protein 1, transforming growth factor beta-activated kinase-binding protein 1, mitogen-activated protein kinase kinase kinase 7 interacting protein 1, Mitogen-activated protein kinase kin

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€381.00

€381.00


Estimated to ship on25 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
€381.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€381.00


Estimated to ship on25 April 2025Details


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat, human, horse, mouse

species reactivity (predicted by homology)

equine (based on 100% sequence homology), ox (based on 100% sequence homology)

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

phosphorylation (pThr431)

Gene Information

human ... TAB1(10454)

General description

TAK1 binding protein 1 (TAB1) is thought to activate TAK1 MAPKKK in transforming growth factor-beta (TGF-beta) signaling. TAB1 can also associate with TAK1 to form a unique MAPKKK complex that is thought to activate AP-1 and the NFkappaB signaling pathways. Through conformational changes, TAB1 is also thought to induce p38alpha autophosphorylation. It has been linked to both IL-1alpha and TNFalpha signaling and therefore may also be involved in the inflammatory response. Recent research has found that the TAK1-TAB1 complex may have anti-inflammatory effects by aiding stercurensin in regulating the NFκB-dependent inflammatory pathways.

Specificity

This antibody recognizes TAB1 phosphorylated at Thr431.

Immunogen

Epitope: Phosphorylated Thr431
KLH-conjugated linear peptide corresponding to human TAB1 phosphorylated at Thr431.

Application

Research Category
Inflammation & Immunology
Research Sub Category
Immunoglobulins & Immunology
This Anti-phospho-TAB1 (Thr431) Antibody is validated for use in WB for the detection of phospho-TAB1 (Thr431).

Quality

Evaluated by Western Blot in L6 cell lysate.

Western Blot Analysis: 1 µg/mL of this antibody detected TAB1 on 10 µg of L6 cell lysate.

Target description

~78 kDa observed. 55 kDa calculated. A higher molecular weight on western blot may be due to glycosylation. An uncharacterized band appears at ~42 and 50 kDa in some lysates.

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
L6 cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bjarte Bergstrøm et al.
Journal of immunology (Baltimore, Md. : 1950), 195(3), 1100-1111 (2015-06-19)
Staphylococcus aureus may cause serious infections and is one of the most lethal and common causes of sepsis. TLR2 has been described as the main pattern recognition receptor that senses S. aureus and elicits production of proinflammatory cytokines via MyD88
Briana Ortiz et al.
American journal of reproductive immunology (New York, N.Y. : 1989), 89(3), e13664-e13664 (2022-12-11)
During gestation, the decidua is an essential layer of the maternal-fetal interface, providing immune support and maintaining inflammatory homeostasis. Although Chlamydia (C.) trachomatis is associated with adverse pregnancy outcomes the pathogenic effects on maternal decidua contributing to adverse events are
Chia-Yu Yang et al.
Journal of immunology (Baltimore, Md. : 1950), 192(4), 1547-1557 (2014-01-10)
T cell activation is dependent upon phosphorylation of MAPKs, which play a critical role in the regulation of immune responses. Dual-specificity phosphatase 14 (DUSP14; also known as MKP6) is classified as a MAPK phosphatase. The in vivo functions of DUSP14
Mariana de Castro Silva et al.
Reproduction (Cambridge, England), 160(4), 627-638 (2020-08-26)
A non-reversible state of epithelial to mesenchymal transition (EMT) at term accumulates proinflammatory mesenchymal cells and predisposes fetal membrane to weakening prior to delivery at term. We investigated the induction of EMT in amnion epithelial cells (AEC) in response to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service