Skip to Content
Merck
All Photos(1)

Key Documents

05-806-AF488

Sigma-Aldrich

Anti-phospho-Histone H3 (Ser10) Antibody, clone 3H10, Alexa Fluor 488

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€446.00

€446.00


Estimated to ship on26 March 2025Details


Request a Bulk Order

Select a Size

Change View
100 μL
€446.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€446.00


Estimated to ship on26 March 2025Details


Request a Bulk Order

conjugate

ALEXA FLUOR 488

Quality Level

clone

3H10
monoclonal

purified by

using protein G

species reactivity

mouse, human

concentration

(Please refer to lot specific datasheet.)

technique(s)

immunocytochemistry: suitable

isotype

IgG1κ

UniProt accession no.

General description

A proprietary immunogen based on a peptide sequence containing phospho-serine corresponding to residue 10 of human histone H3.

Specificity

Histone H3 phosphorylated at Ser10.

Application

Epitope: Histone H3 pSer10.

Quality

Evaluated by Immunocytochemistry in HeLa cells.

Immunocytochemistry Analysis: A 1:100 dilution of this antibody detected histone H3 Ser10 phosphorylation in HeLa cells.

Physical properties

~17 kDa observed.

Physical form

Purified mouse monoclonal IgG1κ antibody conjugate in PBS with 15 mg/mL BSA and 0.1 % sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Human. Broad species cross-reactivity is expected.

Legal Information

ALEXA FLUOR is a trademark of Life Technologies

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service