Skip to Content
Merck
All Photos(1)

Key Documents

05-773

Sigma-Aldrich

Anti-phospho-MYPT1 (Thr850) Antibody, clone SA19, rabbit monoclonal

clone SA19, Upstate®, from rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
clone:
SA19, monoclonal
application:
WB
species reactivity:
human, chicken
technique(s):
western blot: suitable
citations:
7

biological source

rabbit

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

SA19, monoclonal

species reactivity

human, chicken

manufacturer/tradename

Upstate®

technique(s)

western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

phosphorylation (pThr850)

Gene Information

human ... PPP1R12A(4659)

Specificity

MYPT1 phosphorylated on Thr850
Predicted to cross react with mouse and rat

Immunogen

KLH-conjugated, synthetic peptide containing the sequence ...RS[pT]GV..., in which [pT] corresponds to phosphorylated threonine at amino acid residue 850 of chicken myosin phosphatase target subunit 1 (MYPT1). Corresponds to Thr853 in human MYPT1.

Application

Anti-phospho-MYPT1 (Thr850) Antibody, clone SA19 is an antibody against phospho-MYPT1 (Thr850) for use in WB.
Research Category
Signaling
Research Sub Category
Kinases & Phosphatases

Quality

Routinely evaluated by immunoblot on RIPA lysates from HEK293 cells phosphorylated by preincubation with ROKα/ROCK-II, active (Catalog # 14-338).

Target description

doublet at 130kDa

Physical form

Format: Purified
Protein A purified

Storage and Stability

2 years at -20°C from date of shipment

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sustained therapeutic hypercapnia attenuates pulmonary arterial Rho-kinase activity and ameliorates chronic hypoxic pulmonary hypertension in juvenile rats.
Peng, G; Ivanovska, J; Kantores, C; Van Vliet, T; Engelberts, D; Kavanagh, BP; Enomoto et al.
American Journal of Physiology. Heart and Circulatory Physiology null
Role of protein phosphatase type 1 in contractile functions: myosin phosphatase.
Hartshorne, David J, et al.
The Journal of Biological Chemistry, 279, 37211-37214 (2004)
Myosin phosphatase: structure, regulation and function.
Ito, Masaaki, et al.
Molecular and Cellular Biochemistry, 259, 197-209 (2004)
RhoA-mediated Ca2+ sensitization in erectile function.
Wang, Hua, et al.
The Journal of Biological Chemistry, 277, 30614-30621 (2002)
Hypertension and RhoA/Rho-kinase signaling in the vasculature: highlights from the recent literature.
Lee, Dexter L, et al.
Hypertension, 44, 796-799 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service