Skip to Content
Merck
All Photos(1)

Key Documents

05-23-3801

Sigma-Aldrich

Endothelin 3, Human and Rat

Attenuates thrombin-evoked aggregation of human platelets.

Synonym(s):

Endothelin 3, Human and Rat, ET-3, CTCFTYKDKECVYYCHLDIIW

Sign Into View Organizational & Contract Pricing

Select a Size

.5 MG
€585.00

€585.00


Estimated to ship on10 April 2025Details


Request a Bulk Order

Select a Size

Change View
.5 MG
€585.00

About This Item

Empirical Formula (Hill Notation):
C121H168N26O33S4
CAS Number:
Molecular Weight:
2643.04
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.77

€585.00


Estimated to ship on10 April 2025Details


Request a Bulk Order

Quality Level

Assay

≥97% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated

solubility

5% acetic acid: 0.5 mg/mL

shipped in

ambient

storage temp.

−20°C

InChI

1S/C121H168N26O33S4/c1-11-62(7)97(117(175)139-89(121(179)180)49-70-53-126-77-25-17-16-24-75(70)77)145-118(176)98(63(8)12-2)144-112(170)88(52-95(157)158)136-105(163)81(44-60(3)4)131-109(167)86(50-71-54-125-59-127-71)134-113(171)90-56-182-181-55-76(124)101(

InChI key

OQGZWNZGVYLIFX-XKIQDWPYSA-N

General description

Attenuates thrombin-evoked aggregation of human platelets. A potent stimulator of interleukin-6 production by endothelial cell lines. Reported to activate phospholipase C in fibroblasts expressing ETA receptors.

Biochem/physiol Actions

Cell permeable: no
Primary Target
Stimulator of interleukin-6 production by endothelial cell lines
Product does not compete with ATP.
Reversible: no

Packaging

Yes

Sequence

H-Cys¹-Thr-Cys³-Phe-Thr-Tyr-Lys-Asp-Lys-Glu-Cys¹¹-Val-Tyr-Tyr-Cys¹⁵-His-Leu-Asp-Ile-Ile-Trp-OH (disulfide bonds: 1 → 15; 3 → 11)

Physical form

Supplied as a trifluoroacetate salt. Sold on the basis of peptide content.

Preparation Note

Dissolve peptide initially at 8 mg/ml in 80% cold acetic acid, then dilute with cold water to 5% acetic acid and 0.5 mg/ml peptide.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 1 month at -20°C.

Other Notes

Gresser, O., et al. 1996. Biochem. Biophys. Res. Commun.224, 169.
Astarie-Dequeker, C., et al. 1995. Br. J. Pharmacol. 114, 524.
Xin, X., et al. 1995. Endocrinology 136, 132.
Brunner, F., et al. 1992. J. Mol. Cell. Cardiol.24, 1291.
Lamers, J.M., et al. 1992. Mol. Cell. Biochem. 116, 59.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service