Skip to Content
Merck
All Photos(1)

Key Documents

05-22-3838

Sigma-Aldrich

BQ-788, Sodium Salt

Synonym(s):

BQ-788, Sodium Salt, N- cis-2,6-Dimethylpiperidinocarbonyl-L-γ-MeLeu-D-Trp(MeOCO)-D-Nle-OH, Na

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€313.00
5 MG
€1,390.00

€313.00


Estimated to ship on24 March 2025Details



Select a Size

Change View
1 MG
€313.00
5 MG
€1,390.00

About This Item

Empirical Formula (Hill Notation):
C34H50N5NaO7
Molecular Weight:
663.78
UNSPSC Code:
12352200
NACRES:
NA.28

€313.00


Estimated to ship on24 March 2025Details


Assay

≥95% (HPLC)

Quality Level

form

lyophilized solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated

color

white to off-white

solubility

0.01% NH4OH: 1 mg/mL
DMSO: 2 mg/mL
methanol: water (3:1): 2 mg/mL

shipped in

ambient

storage temp.

−20°C

General description

A highly selective and competitive ETB receptor antagonist (IC50 = 1.2 nM).
Highly selective ETB-receptor antagonist (IC50 = 1.2 nM).

Biochem/physiol Actions

Cell permeable: no
Primary Target
ETB-receptor antagonist
Product does not compete with ATP.
Reversible: no
Target IC50: 1.2 nM as ETB-receptor antagonist

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). DMSO and Methano:H₂O stock solutions are stable for up to 2 months at -20°C.

Other Notes

Okada, M., and Nishikibe, M. 2002. Cardiovasc. Drug Reviews20, 53.
Taniai, H., et al. 2001. Hepatology33, 894.
Fukuroda, T., et al. 1994. Biochem. Biophys. Res. Commun.199, 1461.
Ishikawa, K., et al. 1994. Proc. Natl. Acad. Sci. USA91, 4892.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service