Skip to Content
Merck
All Photos(4)

Documents

05-1431

Sigma-Aldrich

Anti-Casein Kinase 2α Antibody, 1AD9

clone 1AD9, from mouse

Synonym(s):

CK2 catalytic subunit alpha, CKII alpha, Casein kinase II subunit alpha, Casein kinase 2, alpha 1 polypeptide, Casein kinase II alpha 1 subunit, Casein kinase II-alpha subunit, Protein kinase CK2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1AD9, monoclonal

species reactivity

human, mouse, rat

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable (paraffin)
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CSNK2A1(1457)

General description

Unlike most serine/threonine kinases, Casein Kinases (CKs) are not activated by specific stimuli, but are essentially constitutively active. CK1 autophosphorylates, causing inactivation, and protein phosphatases reactivate the kinase. The substrates of CKs are numerous, but their function is less clear, perhaps due to the absence of obvious regulation. CK1 isoforms are thought to be involved in the response to DNA damage, while CK2 levels are elevated in many types of cancer.

Specificity

The antibody binds specifically to the catalytic α-subunit of casein kinase2 (CK-2α). The antibody immunoprecipitates a protein with a molecular weight of 44 kD, CK-2α subunit. A 26 kD protein band, the β-subunit of the heterotetrameric holoenzyme α2β2, is coprecipitated with the CK-2α subunit. In western blots with extracts of kidney tissue, the antibody detects various protein bands between 40 and 44 kDa. Using 1 μg/mL of antibody, the lower detection limit for the holoenzyme is 500 ng in Western blot and 100 ng in dot blot; the detection limits for the α-subunit are 300 ng and 60 ng, respectively.

Immunogen

Epitope: Region common to both alpha and alpha′ subunits
Purified recombinant α-subunit of casein kinase 2.

Application

Anti-Casein Kinase 2α Antibody, 1AD9 detects level of Casein Kinase 2α & has been published & validated for use in IC, WB, IH(P) & IP.
Immunoprecipitation: Recommended

Immunocytochemistry: A previous lot of this antibody was used at a dilution of 5 μg/mL (acetone or methanol fix only).
Optimal working dilutions must be determined by end user.

Immunohistochemistry(paraffin): Representative testing from a previous lot.
Optimal Staining of Casein Kinase 2 Monoclonal Antibody: S-KB-3 Cells
Research Category
Signaling

Apoptosis & Cancer
Research Sub Category
Developmental Signaling

Quality

Routinely evaluated by Western Blot on PC12 lysates.

Western Blot Analysis: 1:500 dilution of this lot detected Neurofilament on 10 μg of PC12 lysates.

Target description

44 kDa

Linkage

Replaces: 04-1129

Physical form

Format: Purified
Purified
Purified mouse monoclonal IgG1 in buffer containing 0.02 M Phosphate, pH 7.6 with 0.25 M NaCl and 0.1% sodium azide.

Storage and Stability

Stable at 2-8°C in undiluted aliquots for up to 6 months from date of receipt.

Analysis Note

Control
HeLa cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yoko Yoshihisa et al.
Phytotherapy research : PTR, 34(6), 1320-1328 (2019-12-17)
Ultraviolet (UV) radiation elicits melanogenesis and pigmentation in the skin. Apigenin (4',5,7-trihydroxyflavone [AGN]) is a plant flavone contained in various herbs, fruits, and vegetables. We herein investigated antimelanogenic properties of AGN and the molecular mechanisms of the action of AGN.
Ji-Eun Kim et al.
International journal of molecular sciences, 21(24) (2020-12-23)
Both α-amino-3-hydroxy-5-methylisoxazole-4-propionic acid receptor (AMPAR) and N-methyl-D-aspartate receptor (NMDAR) have been reported as targets for treatment of epilepsy. To investigate the roles and interactions of AMPAR and NMDAR in ictogenesis of epileptic hippocampus, we analyzed AMPAR antagonists (perampanel and GYKI
Yoko Yoshihisa et al.
Phytotherapy research : PTR, 34(6), 1320-1328 (2019-12-17)
Ultraviolet (UV) radiation elicits melanogenesis and pigmentation in the skin. Apigenin (4',5,7-trihydroxyflavone [AGN]) is a plant flavone contained in various herbs, fruits, and vegetables. We herein investigated antimelanogenic properties of AGN and the molecular mechanisms of the action of AGN.
Estefania Alcaraz et al.
PloS one, 15(1), e0227340-e0227340 (2020-01-08)
The PI3K/Akt pathway is interconnected to protein kinase CK2, which directly phosphorylates Akt1 at S129. We have previously found that, in HK-2 renal cells, downregulation of the CK2 regulatory subunit β (shCK2β cells) reduces S129 Akt phosphorylation. Here, we investigated
Jordi Vilardell et al.
Oncotarget, 9(5), 5736-5751 (2018-02-22)
Clear cell renal cell carcinoma (ccRCC) is the most common and aggressive subtype of renal cancer. STAT3 pathway is altered in these tumors and p-STAT3 Ser727 is an independent prognostic factor for ccRCC. Protein kinase CK2 is altered in different

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service