Skip to Content
Merck
All Photos(1)

Documents

05-1317

Sigma-Aldrich

Anti-SUZ12 Antibody, clone 2AO9

clone 2AO9, from mouse

Synonym(s):

ChET 9 protein, Chromatin precipitated E2F target 9 protein, Joined to JAZF1 protein, Suppressor of zeste 12 protein homolog, joined to JAZF1, suppressor of zeste 12 homolog (Drosophila)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2AO9, monoclonal

species reactivity

human

species reactivity (predicted by homology)

rabbit (based on 100% sequence homology)

technique(s)

ChIP: suitable
immunocytochemistry: suitable
immunoprecipitation (IP): suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... SUZ12(23512)

General description

SUZ12 is a polycomb group (PcG) protein. PcG proteins act by forming multiprotein complexes, which are required to maintain the transcriptionally repressive state of homeotic genes throughout development. PcG proteins are not required to initiate repression, but to maintain it during later stages of development. They probably act via the methylation of histones, rendering chromatin heritably changed in its expressibility. This protein is component of the PRC2 complex, which methylates ′Lys-9′ and ′Lys-27′ residues of histone H3.

Specificity

This antibody recognizes SUZ12.

Immunogen

Epitope: Unknown
Recombinant protein corresponding to human SUZ12.

Application

Immunocytochemistry Analysis: A previous lot was used by an independent laboratory in IC. Pasini, D., et al. (2007). Molecular and Cellular Biology. 27(10):3769–3779.

Chromatin Immunoprecipitation Analysis: A previous lot was used by an independent laboratory in ChiP. Pasini, D., et al. (2007). Molecular and Cellular Biology. 27(10):3769–3779.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Chromatin Biology
Use Anti-SUZ12 Antibody, clone 2AO9 (Mouse Monoclonal Antibody) validated in IP, ICC to detect SUZ12 also known as ChET 9 protein, Chromatin precipitated E2F target 9 protein, Joined to JAZF1 protein.

Quality

Evaluated by Immunoprecipitation in HeLa cell lysate.

Immunoprecipitation Analysis: 10 µg/mL of this antibody immunoprecipitated SUZ12 in HeLa cells.

Target description

~ 95 kDa

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG in buffer containing 0.1 M Tris-Glycine (pH 7.4, 150 mM NaCl) with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
HeLa cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dimitris Karamitros et al.
Development (Cambridge, England), 142(1), 70-81 (2014-12-18)
Balancing stem cell self-renewal and initiation of lineage specification programs is essential for the development and homeostasis of the hematopoietic system. We have specifically ablated geminin in the developing murine hematopoietic system and observed profound defects in the generation of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service