Skip to Content
Merck
All Photos(1)

Key Documents

04-894

Sigma-Aldrich

Anti-phospho-KDR/Flk-1/VEGFR2 (Tyr1054) Antibody, clone D1W, rabbit monoclonal

culture supernatant, clone D1W, Upstate®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

culture supernatant

antibody product type

primary antibodies

clone

D1W, monoclonal

species reactivity

mouse, human

manufacturer/tradename

Upstate®

technique(s)

western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

phosphorylation (pTyr1054)

Gene Information

human ... KDR(3791)

Specificity

KDR/Flk-1/VEGFR2 phosphorylated on Tyrosine 1054.
Predicted to have broad species cross-reactivity based on sequence homology.

Immunogen

KLH-conjugated, synthetic peptide encompassing the phosphorylated tyrosine residue 1054 of human KDR/Flk-1/VEGFR2.

Application

Research Category
Signaling
Research Sub Category
Growth Factors & Receptors
This Anti-phospho-KDR/Flk-1/VEGFR2 (Tyr1054) Antibody, clone D1W is validated for use in WB for the detection of phospho-KDR/Flk-1/VEGFR2 (Tyr1054).
Western Blot: 1:1000-1:4000 dilution for the detection of phospho-KDR/Flk-1/VEGFR2 in RIPA lysates from CSF stimulated porcine aortic endothelial (PAE) cells expressing a human CSF-R1/murine KDR chimeric receptor.

Quality

Routinely evaluated by immunoblot in RIPA lysates from CSF stimulated porcine aortic endothelial (PAE) cells expressing a human CSF-R1/murine KDR chimeric receptor.

Target description

152 kDa

Physical form

Cultured supernantant with 0.05% sodium azide

Storage and Stability

2 years at -20°C from date of shipment.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gareth W Fearnley et al.
Biology open, 5(5), 571-583 (2016-04-06)
Vascular endothelial growth factor A (VEGF-A) binding to the receptor tyrosine kinase VEGFR2 triggers multiple signal transduction pathways, which regulate endothelial cell responses that control vascular development. Multiple isoforms of VEGF-A can elicit differential signal transduction and endothelial responses. However
Edward J Hartsough et al.
Science signaling, 6(304), ra104-ra104 (2013-12-05)
Activation of vascular endothelial growth factor receptor-2 (VEGFR-2), an endothelial cell receptor tyrosine kinase, promotes tumor angiogenesis and ocular neovascularization. We report the methylation of VEGFR-2 at multiple Lys and Arg residues, including Lys(1041), a residue that is proximal to
Phosphorylation of Akt and ERK1/2 is required for VEGF-A/VEGFR2-induced proliferation and migration of lymphatic endothelium.
Dellinger, MT; Brekken, RA
Testing null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service