04-1584
Anti-Interleukin-2 (IL-2) Antibody, clone 3F9
clone AB12.38.35C3.3F9, from mouse
Synonym(s):
interleukin 2, involved in regulation of T-cell clonal expansion, aldesleukin, T cell growth factor, lymphokine, Aldesleukin, interleukin-2, T-cell growth factor
About This Item
Recommended Products
biological source
mouse
Quality Level
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
AB12.38.35C3.3F9, monoclonal
species reactivity
human
technique(s)
western blot: suitable
isotype
IgG2bκ
NCBI accession no.
UniProt accession no.
shipped in
wet ice
target post-translational modification
unmodified
Gene Information
human ... IL2(3558)
General description
Immunogen
Application
Inflammation & Immunology
Cytokines & Cytokine Receptors
Quality
Western Blot Analysis: 0.5 µg/mL of this antibody detected 10 µg of human IL-2 recombinant protein.
Target description
Physical form
Storage and Stability
Analysis Note
Human IL-2 recombinant protein
Other Notes
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service