Skip to Content
Merck
All Photos(2)

Key Documents

04-1545

Sigma-Aldrich

Anti-RARα Antibody, clone 9α-9A6

ascites fluid, clone 9α-9A6, from mouse

Synonym(s):

Nuclear receptor subfamily 1 group B member 1, Retinoic acid receptor, alpha polypeptide, nucleophosmin-retinoic acid receptor alpha fusion protein, NPM-RAR long form, retinoic acid receptor, alpha

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€389.00

€389.00


Estimated to ship on24 March 2025Details

A recombinant, preservative-free antibody is available for your target. Try ZRB1369

Request a Bulk Order

Select a Size

Change View
100 μL
€389.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€389.00


Estimated to ship on24 March 2025Details

A recombinant, preservative-free antibody is available for your target. Try ZRB1369

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

ascites fluid

antibody product type

primary antibodies

clone

9α-9A6, monoclonal

species reactivity

human

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... RARA(5914)

General description

Retinoic acid receptor alpha (RARa) is a nuclear receptor encoded by the RARA gene. This gene is sometimes referred to as NR1B1 (nuclear receptor subfamily 1, group B, member 1). RARa is a member of the steroid/thyroid superfamily of nuclear receptor proteins that regulate gene expression by binding to RNA and other elements of DNA response. As implied by its name, this protein is a receptor for retinoic acid, a metabolite of vitamin A. Retinoic acid is necessary for normal organogenesis, but can act as a teratogen when present at high levels during embryonic and fetal development. During fetal and embryonic development retinoic acid regulates transcription through the activation of nuclear receptors, specifically the Retinoic Acid Receptor (RAR). Signaling is transduced by 2 families of nuclear receptors, retinoic acid receptor (RAR) and retinoid X receptor (RXR), which form RXR/RAR heterodimers. There are three isoforms associated with RAR; RARα, RARβ and RARγ.

Specificity

Homology to mouse and rat is 91%.
This antibody recognizes RARα.

Immunogen

Epitope: Unknown
Recombinant protein corresponding to human RARα.

Application

Anti-RARα Antibody, clone 9α-9A6 is a Mouse Monoclonal Antibody for detection of RARα also known as Retinoic acid receptor alpha polypeptide & has been validated in WB, ICC.
Immunocytochemistry Analysis: 1:500 dilution from a representative lot detected RARα in HeLa and A431 cells.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Transcription Factors

Quality

Evaluated by Western Blot in HL60 cell lysate.

Western Blot Analysis: 1:1,000 dilution of this antibody detected RARα on 10 µg of HL60 cell lysate.

Target description

58 kDa observed

Physical form

Unpurified
Unpurified mouse monoclonal IgG1κ ascites with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
HL60 cell lysate

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service