Skip to Content
Merck
All Photos(1)

Key Documents

04-151

Sigma-Aldrich

Anti-IFNAR-1 Antibody, clone MARI-5A3

clone MARI-5A3, from mouse

Synonym(s):

interferon (alpha, beta and omega) receptor 1, beta-type antiviral protein, alpha-type antiviral protein, Type I interferon receptor 1, Cytokine receptor family 2 member 1, interferon-beta receptor 1, Cytokine receptor class-II member 1, IFN-alpha-REC, i

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€415.00

€415.00


Estimated to ship on28 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
€415.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€415.00


Estimated to ship on28 April 2025Details


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

MARI-5A3, monoclonal

species reactivity

mouse

technique(s)

flow cytometry: suitable
immunoprecipitation (IP): suitable
neutralization: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

mouse ... Ifnar1(15975)

General description

Interferon alpha/beta receptor 1 (IFNAR-1) is a single-pass type I membrane protein containing three fibronectin type-III domains and is a member of the type II cytokine receptor family. IFNAR-1 serves as an interferon receptor specific to interferons alpha and beta, a family of type I interferons critical to the promotion and the linking of adaptive and innate immune response. Type I interferons all bind to a heterodimeric receptor composed of two subunits, IFNAR-1 and IFNAR-2. The binding of the type I IFNs initializes tyrosine phosphorylation for several proteins such as; IFNR alpha/beta subunits, JAKs, STAT, and TYK2 proteins.

Specificity

This antibody recognizes the extracellular domain of IFNAR-1.

Immunogen

Recombinant protein corresponding to the extracellular domain of mouse IFNAR-1.

Application

Anti-IFNAR-1 Antibody, clone MARI-5A3 is an antibody against IFNAR-1 for use in FC, IP, WB, NEUT.
Western Blot Analysis: A previous lot was used by an independent laboratory in WB. (Sheehan, K., et al. (2006). Journal of Interferon & Cytokine Research. 26:804-819.)

Immunoprecipitation Analysis: A previous lot was used by an independent laboratory in IP. (Sheehan, K., et al. (2006). Journal of Interferon & Cytokine Research. 26:804-819.)

Flow Cytometry Analysis: A previous lot was used by an independent laboratory in FC. (Sheehan, K., et al. (2006). Journal of Interferon & Cytokine Research. 26:804-819.)

Neutralizing Assay: A previous lot was used by an independent laboratory in neutralization studies. (Sheehan, K., et al. (2006). Journal of Interferon & Cytokine Research. 26:804-819.)

Quality

Evaluated by Flow Cytometry in BALB/c splenocytes.

Flow Cytometry Analysis: 5 µg of this antibody detected IFNAR-1 in BALB/c splenocytes.

Target description

63 kDa calculated

Physical form

Format: Purified

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Louise Malle et al.
Immunity, 55(11), 2074-2084 (2022-10-16)
Down syndrome (DS) is typically caused by triplication of chromosome 21. Phenotypically, DS presents with developmental, neurocognitive, and immune features. Epidemiologically, individuals with DS have less frequent viral infection, but when present, these infections lead to more severe disease. The

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service