Skip to Content
Merck
All Photos(1)

Documents

04-1460

Sigma-Aldrich

Anti-PRMT5 Antibody, clone 23C7

clone 23C7, from mouse

Synonym(s):

72 kDa ICln-binding protein, HMT1 hnRNP methyltransferase-like 5, Jak-binding protein 1, SKB1 homolog, SKB1 homolog (S. pombe), Shk1 kinase-binding protein 1 homolog, protein arginine methyltransferase 5, skb1 (S. pombe) homolog

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

23C7, monoclonal

species reactivity

human

technique(s)

western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PRMT5(10419)

General description

Arginine methylation of proteins is mediated by the Protein-Arginine Methyl Transferase (PRMT) family of enzymes, which are important in signal transduction, transcription, RNA transport, and splicing. PRMTs are divided into two types defined by their activity. Type I PRMTs (including PRMT 1, 3, 4, and 6) are characterized by the formation of asymmetric dimethylated arginine residues. Type II PRMTs, which include only PRMT5, are defined by the formation of symmetric dimethylated arginine residues. Substrates for PRMT5 include myelin basic protein (MBP) and the spliceosomal D1 and D3 proteins. PRMT5 (also known as Skb1Hs/JBP1) exists as homooligomeric complexes, which includes a dimer and tetramer. Homooligomerization of PRMT5 has an important role in its ability to methylate the MBP protein.

Specificity

This antibody recognizes PRMT5.

Immunogen

Recombinant protein corresponding to human PRMT5.

Application

Research Sub Category
Histone Modifying Proteins

RNA Metabolism & Binding Proteins
Use Anti-PRMT5 Antibody, clone 23C7 (mouse monoclonal antibody) validated in WB to detect PRMT5 also known as 72 kDa ICCln-binding protein, HMT1 hnRNP methyltransferase-like 5, Jak-binding protein 1, SKB1 homolog.

Quality

Evaluated by Western Blot in K562 cell lysate.

Western Blot Analysis: 0.5 µg/mL of this antibody detected PRMT5 on 10 µg of K562 cell lysate.

Target description

~72 kDa observed

Linkage

Replaces: MABE551

Physical form

Format: Purified

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service