Skip to Content
Merck
All Photos(1)

Documents

03-34-0002

Sigma-Aldrich

H-Arg-Gly-Asp-Ser-OH

Synonym(s):

H-Arg-Gly-Asp-Ser-OH, Fibronectin Inhibitor, RGDS

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H27N7O8
CAS Number:
Molecular Weight:
433.42
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.77

Assay

≥97% (HPLC)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

color

white to off-white

solubility

water: 1 mg/mL

shipped in

ambient

storage temp.

−20°C

InChI

1S/C15H27N7O8/c16-7(2-1-3-19-15(17)18)12(27)20-5-10(24)21-8(4-11(25)26)13(28)22-9(6-23)14(29)30/h7-9,23H,1-6,16H2,(H,20,27)(H,21,24)(H,22,28)(H,25,26)(H,29,30)(H4,17,18,19)/t7-,8-,9-/m0/s1

InChI key

NNRFRJQMBSBXGO-CIUDSAMLSA-N

General description

Shown to inhibit fibronectin function by binding to platelet-binding sites.

Biochem/physiol Actions

Cell permeable: no
Primary Target
fibronectin
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Standard Handling (A)

Physical form

Supplied as a trifluoroacetate salt.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Perlot, R.L., et al. 2002. J. Bone Miner. Res.17, 66.
Ginsberg, M., et al. 1985. J. Biol. Chem.260, 3931.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service