Skip to Content
Merck
All Photos(1)

Key Documents

03-209

Sigma-Aldrich

SET7/9 human recombinant - Methyltransferase enzyme

Human recombinant SET7/9 expressed in E. Coli.

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€573.00

€573.00


Estimated to ship on11 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
€573.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.54

€573.00


Estimated to ship on11 April 2025Details


Request a Bulk Order

biological source

human

Quality Level

recombinant

expressed in E. coli

Assay

>95% (SDS-PAGE)

mol wt

~41 kDa

packaging

pkg of 100 μg

manufacturer/tradename

Upstate®

storage condition

avoid repeated freeze/thaw cycles

concentration

1 mg/mL

technique(s)

enzyme immunoassay: suitable

UniProt accession no.

General description

The methylation of histones plays a pivotal role in the regulation of chromatin structure and gene expression. Histone methylation can occur on Arg or Lys residues, with an exquisite site selectivity for Lys methylation at specific positions in the N-termini of histones H3 and H4. SET7/9, a histone methyltransferase (HMTase), which transfers methyl groups to Lys4 of Histone H3, forms a complex with S-adenosyl-L-methionine. This complex contains an active site consisting of a binding pocket where an AdoMet molecule, in an unusual conformation, binds a narrow substrate-specific channel that only unmethylated lysine residues can access, and a catalytic tyrosine residue.

Specificity

<1.0EU/µg protein (LAL test)

Application

Human recombinant SET7/9 expressed in E. Coli.

Physical form

Format: Purified
In liquid containing 50 mM Tris-HCl, pH 7.5, containing 0.2 M sodium chloride, 5 mM DTT and 20% glycerol. Frozen solution.

Storage and Stability

Stable for 12 months at -20°C from date of receipt. Avoid multiple freeze/thaw cycles.

Analysis Note

Methyltransferase assays

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service