Skip to Content
Merck
All Photos(1)

Key Documents

01-195

Sigma-Aldrich

CNTF, recombinant

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
€631.00

€631.00


Estimated to ship on11 April 2025Details



Select a Size

Change View
25 μG
€631.00

About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.41

€631.00


Estimated to ship on11 April 2025Details


biological source

rat

Quality Level

manufacturer/tradename

Upstate®

technique(s)

cell culture | mammalian: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

Gene Information

rat ... CNTF(25707)

General description

Product Source: Recombinant rat Ciliary Neurotrophic Factor expressed in E. coli

Quality

Routinely evaluated in a biological activity assay.

Physical form

Lyophilized from 5mM sodium acetate, pH 6.5.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Prabakaran Soundararajan et al.
Stem cells (Dayton, Ohio), 25(7), 1697-1706 (2007-03-31)
Directing embryonic stem (ES) cells to differentiate into functional motoneurons has proven to be a strong technique for studying neuronal development as well as being a potential source of tissue for cell replacement therapies involving spinal cord disorders. Unfortunately, one

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service