01-125-M
Nerve Growth Factor 2.5s mouse
recombinant, expressed in male mouse submaxillary glands, suitable for cell culture
Synonym(s):
NGF-2.5S
Select a Size
About This Item
Recommended Products
Product Name
NGF 2.5S Protein, mouse,
recombinant
expressed in (Male mouse submaxillary glands)
Quality Level
100
300
purified by
filtration through 0.2μm medium
technique(s)
cell culture | mammalian: suitable
UniProt accession no.
Gene Information
mouse ... NTF3(18205)
General description
SIZE: 257 amino acids; 29355 Da
SUBCELLULAR LOCATION: Secreted.
TISSUE SPECIFICITY: Brain and peripheral tissues.
SIMILARITY: SwissProt: P20783 ## Belongs to the NGF-beta family.
Quality
Physical form
Disclaimer
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service